Product Image
Applied Biosystems™

PharmacoScan™ Reagent Kit, 96-format

The Applied Biosystems PharmacoScan Reagent Kit provides standalone reagents and controls for use with PharmacoScan array plates.
Have Questions?
Catalog NumberQuantity
91302594 samples
Catalog number 913025
Price (USD)
-
Quantity:
94 samples

The Applied Biosystems PharmacoScan Reagent Kit provides standalone reagents and controls for use with PharmacoScan array plates. Each kit includes sufficient reagents and controls to process 94 samples and 2 controls. (2 x 96-format array plates; 22 samples + 2 controls each plate).

Note: These reagents are included in the 96-format PharmacoScan Assay Kit and the 96-format PharmacoScan Training Kit.

PharmacoScan Solution is an ideal platform for translational researchers in healthcare, pharma, and research organizations looking to understand how genetic variations affect drug absorption, distribution, metabolism, and excretion (ADME). By pre-emptively screening for risk factors on a microarray, users in research can precisely and reproducibly analyze 4,627 high- to low-evidence markers within nearly 1,200 genes in a single assay. The solution includes copy number variation calling and interrogating predictive markers in complex genes. Unlike solutions that analyze only high-evidence markers, PharmacoScan Solution presents a more comprehensive view for genotyping for pharmacogenomic risk.

Related Links:

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Application)PharmacoScan Solution workflow (PGx Microarray applications)
IncludesReagent
No. of Samples94 samples
Product LinePharmacoScan
Quantity94 samples
Reagent TypeControls, Primer/Probes, Wash Buffer
TypeReagent Kit
Format96-array Plate
Unit SizeEach
Contents & Storage
1 reagent kit (includes reagents and controls for 1 x 96-format array plate)

Frequently asked questions (FAQs)

Do you offer an alternative for the discontinued DMET PLUS Assay (Cat. No. 901268)?

We recommend reviewing the Microarrays for Preemptive Pharmacogenomics page to determine a potential alternative.

If you have any questions you can email Tech Support at techsupport@thermofisher.com.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.