BacMam pCMV-Dest Vector
Invitrogen™

BacMam pCMV-Dest Vector

The BacMam pCMV-Dest Vector combines Gateway™ cloning and BacMam gene expression technologies for easy recombination-based cloning and baculovirus-based expression of a target gene in variety of cell types.
Have Questions?
Catalog NumberQuantity
A2422310 μg
Catalog number A24223
Price (USD)
1,090.00
Each
Quantity:
10 μg
Price (USD)
1,090.00
Each

The BacMam pCMV-Dest Vector combines Gateway™ cloning and BacMam gene expression technologies for easy recombination-based cloning and baculovirus-based expression of a target gene in variety of cell types. With BacMam technology, a modified insect cell virus (baculovirus) is used as a vehicle to efficiently deliver and transiently express genes in mammalian cells with minimum effort and toxicity. This technology allows for control over the level of expression through the increase or decrease of viral particle concentration. When combined with Gateway™ cloning technology, a variety of sizes of target genes can be cloned and expressed. Sizes ranging from an insert for RNAi to a large gene with introns (38 kb) have been cloned and expressed successfully using the BacMam pCMV-Dest Vector. This plasmid accommodates various cloning schemes, including Gateway™, Seamless Assembly, and traditional cloning using restriction enzymes.

The BacMam pCMV-Dest Vector contains a CMV promoter to drive constitutive expression of the target gene. It also contains VSV-G elements, which enable baculovirus to have high transduction efficiency, and a WPRE element, which elongates transient expression in mammalian systems. To facilitate successful cloning, AmpR and gentamicin have been incorporated for positive selection, as well as Cm(R) (chloramphenicol resistance gene) for negative selection. Upon successful cloning, the region containing the CmR is replaced by the insert, selecting against any bacteria that still have a CmR-containing region.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Delivery TypeBaculovirus (BacMam System)
For Use With (Application)Viral Expression, Constitutive Expression, Multi-Gene Expression
Product TypeInsect Cell Expression Vector
Quantity10 μg
Reporter GeneNone
Selection Agent (Eukaryotic)None
VectorpDEST
Cloning MethodRestriction Enzyme/MCS, Seamless Cloning, Gateway
Concentration500 ng/μL
Product LineViraPower
PromoterCMV
Protein TagUntagged
Unit SizeEach
Contents & Storage
10 μg of BacMam pCMV-Dest Vector.
Store at 2°C to 8°C.

Citations & References (1)

Citations & References
Abstract
Combined TRPC3 and TRPC6 blockade by selective small-molecule or genetic deletion inhibits pathological cardiac hypertrophy.
Authors:Seo K, Rainer PP, Shalkey Hahn V, Lee DI, Jo SH, Andersen A, Liu T, Xu X, Willette RN, Lepore JJ, Marino JP, Birnbaumer L, Schnackenberg CG, Kass DA,
Journal:
PubMed ID:24453217
Chronic neurohormonal and mechanical stresses are central features of heart disease. Increasing evidence supports a role for the transient receptor potential canonical channels TRPC3 and TRPC6 in this pathophysiology. Channel expression for both is normally very low but is increased by cardiac disease, and genetic gain- or loss-of-function studies support ... More