TaqMan™ Advanced miRNA Human Serum/Plasma Card
Applied Biosystems™

TaqMan™ Advanced miRNA Human Serum/Plasma Card

The Applied Biosystems TaqMan Advanced Serum/Plasma Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidicRead more
Have Questions?
Catalog NumberQuantity
A34717384 reactions
Catalog number A34717
Price (USD)
554.00
Each
Quantity:
384 reactions
Price (USD)
554.00
Each
The Applied Biosystems TaqMan Advanced Serum/Plasma Card provides the convenience of pre-spotted TaqMan Advanced miRNA assays in a 384-well microfluidic card format. This product contains one card for profiling of up to 188 unique miRNAs in one serum/plasma sample, and includes endogenous and exogenous miRNA controls for normalization of data results.

Features of the TaqMan Advanced Serum/Plasma Card include:
• Reproducibility—pre-spotted assays are designed to reduce pipetting error and minimize experimental variability
• Sample savings—requires only 2 μL of purified plasma/serum or 1–10 ng of total RNA per sample
• Updated content—assay content updated for mirBase v22
• Easy to use—just add a mix of diluted miR-Amp and master mix and run the qPCR; no additional pipetting required
• Gold-standard TaqMan qPCR chemistry—TaqMan Advanced miRNA assays draw on Thermo Fisher Scientific's industry-leading bioinformatics assay design pipeline and verification system to help ensure high specificity and minimal cross-reactivity, even for closely-related miRNA families

Product details

Number of cards: 1
Number of unique miRNAs (not including exogenous controls): 188
Exogenous controls: cel-miR-39-3p, ath-miR159a
Endogenous control: hsa-miR-16-5p
Samples per card: 1
Compatible with the following Applied Biosystems qPCR instruments: Viia7, QuantStudio 7 Flex, and QuantStudio 12k Flex
Recommended master mix: TaqMan Fast Advanced Master Mix

Notes
• TaqMan Advanced miRNA plates require the use of the TaqMan Advanced cDNA Synthesis Kit, which uses a universal reverse transcription (RT) chemistry to prepare the cDNA template for use with TaqMan Advanced miRNA assays for detection and quantification of mature human miRNAs in biological samples.
• These plates are not compatible with the first-generation TaqMan MicroRNA cDNA Synthesis Kit, which uses an miRNA-specific reverse transcription chemistry.
• For more information on the availability and pricing for Advanced miRNA assays in pre-spotted 384 well plates, please email specialty_cards@thermofisher.com.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Format384-well microfluidic card
Product LineTaqMan
Product TypeAdvanced miRNA Human Serum/Plasma Card
Quantity384 reactions
Research CategorymiRNA
Shipping ConditionApproved for shipment at Room Temperature or on Wet or Dry Ice
SpeciesHuman
Volume2 μL
Detection MethodPrimer-probe
For Use With (Application)miRNA analysis
GC-Rich PCR PerformanceHigh
Label or Dye2 Color
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
One 384-well array

The TaqMan miRNA card is shipped at ambient temperature. Upon arrival, store at 2–8°C and keep protected from light.

Frequently asked questions (FAQs)

I am not seeing good amplification with my TaqMan Advanced miRNA array cards. What is wrong?

The TaqMan Advanced miRNA array cards allow one to profile over 300 different miRNA targets on one card. Due to the large number of targets and the nature of your sample, it is possible that some miRNA on the card will not be expressed on your sample, or expressed at very low levels. This is one reason why you will not see good amplification plots for every spot on the array card. Another possibility is the sample quality or input. Make sure to check the concentration and quality of the RNA sample before reverse transcription. Check the Ct values for your endogenous control assays on the card, and see if it is in the expected range for your typical samples.