Pierce™ Sulfo-SANPAH, No-Weigh™ Format
Thermo Scientific™
Pierce™ Sulfo-SANPAH, No-Weigh™ Format
Thermo Scientific Pierce Sulfo-SANPAH is a hetero-bifunctional crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A3539510 x 1 mg
2258950 mg
Catalog number A35395
Price (USD)
259.00
Each
Quantity:
10 x 1 mg
Price (USD)
259.00
Each
Thermo Scientific Pierce Sulfo-SANPAH is a hetero-bifunctional crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide. NHS esters react efficiently with primary amino groups (-NH2) in pH 7–9 buffers to form stable amide bonds. The reaction results in the release of sulfo-N-hydroxy-succinimide. When exposed to UV light, nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines). Sulfo-SANPAH, supplied as a sodium salt, is useful for cell-surface protein crosslinking and possesses charged groups for water solubility to a concentration of 10 mM.
Thermo Scientific No-Weigh products are specialty reagents provided in a pre-aliquoted format. The pre-weighed packaging prevents the loss of reagent reactivity and contamination over time by eliminating the repetitive opening and closing of the vial. The format enables use of a fresh vial of reagent each time, eliminating the hassle of weighing small amounts of reagents and reducing concerns over reagent stability.
Features of Sulfo-SANPAH:
• Reactive groups: sulfo-NHS ester and nitrophenyl azide
• Reactive towards: amino groups and any nucleophile
• Non-cleavable
• N-Sulfosuccinimidyl-6-(4'-azido-2'-nitrophenylamino) hexanoate
• Optimal photolysis occurs at 320-350 nm; minimizes damage to biomolecules by irradiation
• Water soluble; non-cleavable
Product References:
Crosslinker Application Guide -- search for recent literature references for this product
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityNo
FormPowder
Labeling MethodChemical Labeling
Molecular Weight (g/mol)492.4
PEGylatedNo
Product LineNo-Weigh, Pierce
Quantity10 x 1 mg
Reactive MoietySulfo-NHS Ester, Nitrophenyl Azide
Shipping ConditionAmbient
SolubilityWater
Spacer Arm Length18.2 Å
Water SolubleYes
Chemical ReactivityAmine-Nonselective
CleavableBy Thiols
Crosslinker TypeHeterobifunctional
FormatStandard, Single-use
Product TypeCrosslinker
SpacerMedium (10 to 30 Å)
Unit SizeEach
Contents & Storage
Upon receipt store at -20°C protected from light and moisture.
Frequently asked questions (FAQs)
Can you provide the shelf-life for Sulfo-SANPAH?
Sulfo-SANPAH is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What are the reactive groups on Sulfo-SANPAH and which groups do they react with?
Sulfo-SANPAH is a heterobifunctional crosslinker that has a sulfo-NHS ester and a photoactivatable nitrophenyl azide group. The sulfo-NHS ester reacts with primary amines in pH 7-9 buffers to form stable amide bonds. When exposed to UV light, nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.