High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
High Select™ Depletion Spin Columns
Thermo Scientific™

High Select™ Depletion Spin Columns

Decrease albumin, immunoglobulins, and other high-abundance proteins from human serum or plasma in preparation for mass spectrometry analysis, or 1D and 2D gel electrophoresis using High Select depletion spin columns or resin.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct TypePurification Target
A3636624 ColumnsMini Spin ColumnHSA/Immunoglobulin
A363656 ColumnsMini Spin ColumnHSA/Immunoglobulin
A3636710 ColumnsMidi Spin ColumnHSA/Immunoglobulin
A3636850 mLResinHSA/Immunoglobulin
A3637110 ColumnsMidi Spin ColumnTop14 Abundant Protein
A3637250 mLResinTop14 Abundant Protein
A363696 ColumnsMini Spin ColumnTop14 Abundant Protein
A3637024 ColumnsMini Spin ColumnTop14 Abundant Protein
Catalog number A36366
Price (USD)
713.65
Online exclusive
802.00
Save 88.35
Each
Quantity:
24 Columns
Product Type:
Mini Spin Column
Purification Target:
HSA/Immunoglobulin
Price (USD)
713.65
Online exclusive
802.00
Save 88.35
Each
Analysis of human fluids is often complicated by the presence of high concentrations of serum proteins, including albumin and IgG that can make up more than 70% of total serum protein. Removal of albumin, immunoglobulins, and other abundant proteins is essential for the study of low-abundance proteins. Thermo Scientific High Select HSA/Immunoglobulin and Top 14 Abundant Protein depletion spin columns and resins enable depletion of the most abundant proteins in human serum and plasma for improved detection of low-abundance proteins using mass spectrometry analysis or 1D or 2D gel electrophoresis.
• Flexible—resin provided in mini and midi column formats as well as bulk for custom filling options
• Optimized—removes >95% of IgG and >95% of albumin (HSA/Immunoglobulin); removes >95% of IgG and >95% of albumin, plus 12 other abundant proteins (Top 14)
• Fast—process samples in ∼10 minutes
• Economical—cost-effective spin columns are priced for single use
• Reduced carry-over—one-time use prevents protein carryover and experimental variability

High Select HSA/Immunoglobulin Depletion Resin uses highly specific, immobilized anti-HSA and anti-immuoglobulin antibodies (IgG, IgA, IgM, IgD, and IgE) to remove human serum albumin (HSA) and all major subclasses of immunoglobulins from serum and plasma. High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resins are provided in an economical and convenient spin column format specifically designed for one-step processing and single use or bulk format for custom filling.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeProtein
For Use With (Equipment)Mass Spectrometer
Label or DyeUnlabeled
Purification TargetHSA/Immunoglobulin
Quantity24 Columns
Shipping ConditionWet Ice
Workflow StepProtein Depletion
FormatMini Spin Column
Product LineHigh Select
Product TypeMini Spin Column
Starting MaterialPlasma, Serum
Unit SizeEach
Contents & Storage
Store at 2–8°C.

Frequently asked questions (FAQs)

After depletion of abundant proteins from my sample, I still see albumin and other abundant proteins after LC-MS analysis. Did the depletion work?

Depletion does not remove all abundant proteins from the sample. Even with 99% depletion of a specific plasma protein, highly abundant proteins will still be the most abundant proteins in the sample detected by MS analysis. However, depletion significantly decreases the most abundant plasma proteins enabling identification of lower abundant proteins.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

How do I proceed to downstream mass spec sample preparation after abundant plasma protein depletion?

After the depletion, samples are significantly diluted in buffer. Depleted samples should be concentrated by speed vac drying, solvent precipitation, or diafiltration. Samples dried using a speedvac are directly compatible with the EasyPep kit chemistry after resuspension using the Lysis Solution.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

What is the maximum loading capacity for the High Select Depletion columns?

Mini columns can deplete up to 10 µL (600 µg) of plasma or serum; midi columns can deplete up to 100 µL (6000 µg) of plasma or serum.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

What is the percentage of albumin that is removed using abundant protein depletion columns?

More than 99% of albumin can be depleted when columns use the recommended sample to resin ratio.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.

How do I assess efficiency and reproducibility of abundant protein depletion in plasma samples?

ELISA and mass spectrometry analysis are often used for evaluation of depletion performance. Protein yields after depletion can be measured using the BCA assay.

Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.