SimpliAmp™ Thermal Cycler + Extended Warranty + Plastics Packages
Applied Biosystems™

SimpliAmp™ Thermal Cycler + Extended Warranty + Plastics Packages

The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWarranty
A369695 years of total service coverage with instrument rapid exchange
A369705 years of total service coverage
Catalog number A36969
Price (USD)
7,453.19
Each
Warranty:
5 years of total service coverage with instrument rapid exchange
Price (USD)
7,453.19
Each

The Applied Biosystems SimpliAmp Thermal Cycler is an easy-to use, compact, and accurate thermal cycler designed to fit every lab’s essential PCR workflow. Innovations like a responsive color touch-screen and Applied Biosystems VeriFlex temperature control technology enable simple, accurate optimization.

This package consists of a SimpliAmp Thermal Cycler, plastics consumables, and an extended warranty. You can access your SimpliAmp cycler with Thermo Fisher Connect, giving you the freedom to design and securely upload your methods from any mobile device or desktop computer.

Rapid Exchange (REX) extended warranty

With the REX extended warranty, if your instrument needs repair, just call our Support Center and we will exchange it for a factory-certified refurbished replacement instrument within one business day from the time we determine a replacement is required. Instruments are typically received within three days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.

The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.

AB Repair Center (ABRC) extended warranty

With ABRC support, factory-trained engineers will perform maintenance or repair your instrument at our remote service center. Just send us your instrument and we will maintain or repair it and ship it back to you typically within three weeks. All parts, labor, and shipping charges are included.

The ABRC extended warranty is ideal for customers in regulated environments that must maintain asset tag consistency to comply with regulated protocols.

Contact your Service and Support Representative for details at service.sales@thermofisher.com.

MicroAmp plastics

This package contains several MicroAmp plastic consumables to get you started. All of our plastic consumables are validated with Applied Biosystems instruments for optimal fit and performance. The following plastic consumables will be delivered along with your instrument when you order this package:

  • 1 x MicroAmp 8-Tube Strip with attached domed caps, 0.2 mL (Cat. No. A30589)
  • 1 x MicroAmp Clear Adhesive Film (Cat. No. 4306311)
  • 5 x MicroAmp TriFlex 3 x 32-Well PCR Reaction Plates (Cat. No. A32811)
  • 1 x MicroAmp 32-Well Clear Adhesive Film (Cat. No. A32812)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Dimensions18.1 x 9.5 x 8.3 in. (46 x 24 x 21 cm)
Display Type8 in. Color TFT LCD Touchscreen
High-throughput CompatibilityManual Protocols
Max. Ramp Rate4°C/sec. (Block), 3°C/sec. (Sample)
Quantity1 Package
Reaction SpeedFast, Standard
Thermal Range0°C to 100°C
User InterfaceUSB, Wi-Fi, Ethernet
Voltage100/240 V
Warranty5 years of total service coverage with instrument rapid exchange
Weight18.3 lb. (8.5 kg)
Block Configurations3 x 32-well Isothermal Blocks
Capacity96-well, 0.2 mL
Product TypeThermal Cycler
Unit SizeEach
Package Components
SimpliAmp™ Thermal Cycler
Catalog Number
A24811
Unit Size
1 instrument
Qty
1
SimpliAmp™ Thermal Cycler, 3 years extended warranty, Rapid Exchange
Catalog Number
ZGEXESCSIMPLIAMP3Y
Unit Size
3 yr ext warranty, Rapid Exchange
Qty
1
MicroAmp™ 8-Tube Strip with Attached Domed Caps, 0.2 mL
Catalog Number
A30589
Unit Size
125 strips
Qty
1
MicroAmp™ Clear Adhesive Film
Catalog Number
4306311
Unit Size
100 films
Qty
1
MicroAmp™ TriFlex 3 x 32-Well PCR Reaction Plate
Catalog Number
A32811
Unit Size
20 plates
Qty
5
MicroAmp™ 32-Well Clear Adhesive Film
Catalog Number
A32812
Unit Size
100 films
Qty
1
Contents & Storage
• SimpliAmp Thermal Cycler
• 3-year Rapid Exchange (REX) extended warranty
• 5 x MicroAmp EnduraPlate Optical 96-well Clear Reaction Plates with Barcode (20 plates/pack, 100 plates total)
• 1 x MicroAmp 8-Tube Strip with Attached Domed Caps, 0.2 mL (125 strips)
• 1 x MicroAmp Clear Adhesive Film (100 films)
• 5 x MicroAmp TriFlex 3 x 32-well PCR Reaction Plates (20 plates/pack, 100 plates total)
• 1 x MicroAmp 32-well Clear Adhesive Film (100 films)

Frequently asked questions (FAQs)

What is the optimal ramp rate for my thermal cycler?

There is no universal ramp rate for a thermal cycler because both the block and sample ramp rates are different for each thermal cycler. The sample ramp rate changes with the volume of the sample since more time and energy are required to heat higher sample volumes. The ramp rate may also need to be changed for a particular PCR assay or primer pair if you are trying to use the same PCR assay on two different thermal cyclers, for example.

As a reference, the block ramp rate commonly used with DreamTaq Polymerase on the MiniAmp thermal cycler is 3.0 C/sec. In a 50 µL PCR reaction, this translates to a sample ramp rate of ~1 C/sec.

Please refer to the following application note for further information about thermocycler ramp rates: https://assets.thermofisher.com/TFS-Assets/LSG/brochures/Thermal_Cycler_Ramp_RatesAppNote.pdf

Find additional tips, troubleshooting help, and resources within ourPCR Thermal Cyclers Support Center.