PNGase F Glycan Cleavage Kit
Gibco™

PNGase F Glycan Cleavage Kit

The PNGase F Glycan Cleavage Kit includes all components necessary to perform the enzymatic removal of almost all N-linked oligosaccharides자세히 알아보기
Have Questions?
카탈로그 번호수량
A392451 kit
카탈로그 번호 A39245
제품 가격(KRW)
461,000
온라인 행사
Ends: 30-Jun-2026
526,000
할인액 65,000 (12%)
Each
수량:
1 kit
제품 가격(KRW)
461,000
온라인 행사
Ends: 30-Jun-2026
526,000
할인액 65,000 (12%)
Each
The PNGase F Glycan Cleavage Kit includes all components necessary to perform the enzymatic removal of almost all N-linked oligosaccharides from glycoproteins. The kit includes the PNGase F enzyme, which cleaves N-glycan chains at the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides.

The PNGase F Glycan Cleavage Kit features include:
• ≥95% purity
• Recombinant PNGase F enzyme with no endoglycosidase F1, F2, or F3
• Enzyme leaves N-glycan core oligosaccharides intact and suitable for downstream analysis
• Suitable for use under native or denaturing conditions

The components included in the PNGase F Glycan Cleavage Kit are: 30,000 units of PNGase F (500,000 units/mL) and 1 mL PNGase F 10X buffer.

For Research Use Only. Not for use in diagnostic procedures.
사양
형태Liquid
수량1 kit
배송 조건Wet Ice
제품 유형PNGase F Glycan Cleavage Kit
Unit SizeEach
구성 및 보관
• 30,000 units PNGase F enzyme, recombinant (500,000 units/mL), store at -20°C
• 1 mL PNGase F 10X Buffer, store at -20°C

자주 묻는 질문(FAQ)

In the PNGase Glycan Cleavage Kit, can I use PNGase F under native or denaturing conditions?

Yes, PNGase F enzyme may be used in both native and denaturing conditions

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

In the PNGase Glycan Cleavage Kit, how much PNGase F should I use to remove my carbohydrate under native or DTT denaturing conditions?

When the protein is not denatured with SDS, PNGase F has to work harder to reach the cleavage site of the carbohydrate (due to secondary and tertiary protein structures). In these instances, using more enzyme and/or extended incubation times may help. These conditions will need to be determined empirically for your specific protein.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

In the PNGase Glycan Cleavage Kit, is PNGase F compatible with downstream analysis such as HPLC and mass spectrometry?

PNGase F enzyme is stored in 50% glycerol. Therefore, HPLC and/or mass spectrometry is not compatible due to the fact that glycerol is not tolerated in instruments used for these applications.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

In the PNGase Glycan Cleavage Kit, what are the typical reaction conditions for PNGase F, Recombinant?

Typical reaction conditions are as follows:

Optimal incubation times and enzyme concentrations must be determined empirically for a particular substrate.
1. Combine 1-20 µg of glycoprotein, 1 µL of 10X denaturing buffer (5% SDS, 400 mM DTT) and H2O (if necessary) to make a 10 µL total reaction volume.
2. Denature glycoprotein by heating reaction at 100 degrees C for 10 min.
3. Make a total reaction volume of 20 µL by adding 2 µL PNGaseF 10X Buffer, 1-5 µL PNGaseF enzyme, 2 µL 10% NP-40 and H2O.
4. Incubate reaction at 37 degrees C for 1 hr.
Note: Reactions may be scaled-up linearly to accommodate larger reaction volumes.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

In the PNGase Glycan Cleavage Kit, what happens to the asparagine residue after PNGase F removes the sugar?

As PNGase F is a glycoamidase, the asparagine gets converted into aspartic acid.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.