ReadyCount™ Blue Nuclear Stain, 100 Assays - FAQs
ReadyCount™ Blue Nuclear Stain, 100 Assays - FAQs
View additional product information for ReadyCount™ Stains - FAQs (A49903, A49904, A49905)
5 product FAQs found
How can I use my Countess 3 Automated Cell Counter, for cell viability assays, and what are the steps?
Invitrogen ReadyCount Stains are pre-optimized for viability analysis with Countess 3 FL Automated Cell Counter. Simply mix 10 µL of cell sample with 10 µL of stain, pipet the solution into a Countess chamber or reusable slide and insert the slide into the instrument.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
Which staining dyes can I use with my Countess 3 FL Automated Cell Counter?
EVOS light cubes, which enable fluorescent detection in the Countess 3 FL Automated Cell Counter, are compatible with a broad range of fluorescent dyes. Refer to our selection guide for an extensive list of compatible dyes. We also offer the ready-to-use ReadyCount Stains for easy viability staining.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
What is the expiration date/warranty period for ReadyCount (Cat. No. A49904) Blue Nuclear Stain?
If an expiration date or warranty period is not posted on any public-facing document, the ReadyCount reagents have the standard warranty period of 1 year from the date of shipment.
What is the buffer/solution of the ReadyCount Blue Nuclear Stain (Cat. No. A49904)?
The formulation/identity of components in the solution of ReadyCount Blue Nuclear Stain (Cat. No. A49904) is proprietary.
What is the concentration of the Hoechst in ReadyCount Blue Nuclear Stain (Cat. No. A49904)?
The concentration of Hoechst 33342 in ReadyCount Blue Nuclear Stain (Cat. No. A49904) is proprietary. If you need to use a defined concentration of Hoechst 33342, we do offer this as a standalone product (Cat. Nos. 62249, H1399, and H21492).