Search
View additional product information for ReadyCount™ Stains - FAQs (A49903, A49904, A49905)
5 product FAQs found
Invitrogen ReadyCount Stains are pre-optimized for viability analysis with Countess 3 FL Automated Cell Counter. Simply mix 10 µL of cell sample with 10 µL of stain, pipet the solution into a Countess chamber or reusable slide and insert the slide into the instrument.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
EVOS light cubes, which enable fluorescent detection in the Countess 3 FL Automated Cell Counter, are compatible with a broad range of fluorescent dyes. Refer to our selection guide for an extensive list of compatible dyes. We also offer the ready-to-use ReadyCount Stains for easy viability staining.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
If an expiration date or warranty period is not posted on any public-facing document, the ReadyCount reagents have the standard warranty period of 1 year from the date of shipment.
The formulation/identity of components in the solution of ReadyCount Green/Red Viability Stain (Cat. No. A49905) is proprietary.
The concentrations of Acridine Orange and Propidium iodide in ReadyCount Green/Red Viability Stain (Cat. No. A49905) are proprietary. If you need to use a defined concentration of Acridine Orange, we do offer this as a standalone product (Cat. Nos. A1301 and A3568). If you need to use a defined concentration of Propidium iodide, we do offer this as a standalone product (Cat. Nos. P3566, P1304MP, and P21493).