Absolute Q™ 1-step RT-dPCR Master Mix (4X) - FAQs
View additional product information for Absolute Q™ 1-step RT-dPCR Master Mix (4X) - FAQs (A55146)
1 product FAQs found
Can I use TaqMan SNP Genotyping Assays to detect single-nucleotide variants (SNVs) that are present in less than 5% of a sample?
TaqMan SNP Genotyping Assays are designed to work on equal volumes, so if the mutation is only present at 5%, it is possible that the assay will not detect the variants. Therefore, we do not recommend using these assays to detect somatic mutations. SNP assays by qPCR with TaqMan SNP Genotyping Assays are intended to detect germline mutations.
For somatic mutations, we recommend using digital PCR or castPCR. Find more information about digital PCR and castPCR in the links below:
Digital PCR Assays and Kits
QuantStudio Absolute Q Digital PCR System
Absolute Q 1-Step RT-dPCR Master Mix (4X) (Cat. No. A55146) User Guide
TaqMan Mutation Detection Assay with castPCR Technology (Cat. Nos. 4465804, 4465807)
TaqMan Mutation Detection Assay: Competitive Allele-Specific TaqMan PCR
Find additional tips, troubleshooting help, and resources within our TaqMan Primers and Probes Support Center.