Image-iT™ Fixation/Permeabilization Kit
Invitrogen™

Image-iT™ Fixation/Permeabilization Kit

The Image-iT Fixation/Permeabilization Kit is a convenient-to-use fixation/permeabilization kit that contains all the necessary reagents to prepare cells for antibodyRead more
Have Questions?
Catalog NumberQuantity
A58181011 Kit
Catalog number A5818101
Price (USD)
389.50
Special offer
Online exclusive
Ends: 31-Dec-2026
475.00
Save 85.50
Each
Quantity:
1 Kit
Price (USD)
389.50
Special offer
Online exclusive
Ends: 31-Dec-2026
475.00
Save 85.50
Each

The Image-iT Fixation/Permeabilization Kit is a convenient-to-use fixation/permeabilization kit that contains all the necessary reagents to prepare cells for antibody staining and imaging. The high-quality components help preserve cell morphology and reduce background staining and are ideal for use with antibody-based detection methods, including immunocytochemistry (ICC), immunofluorescence (IF), and immunohistochemistry (IHC). An easy-to-follow protocol is also provided. Note: the 4% formaldehyde, methanol-free fixative solution included in this kit is also available for separate purchase (see Cat. No. R37814).

Features of the Image-iT Fixation/Permeabilization Kit include:
• A complete fixation/permeabilization kit with ready-to-use components and room temperature storage
• Optimized for secondary antibody labeling, fixed-cell dye staining, and GFP applications
• Reduced background staining

See other ReadyProbes reagents for cell staining ›
Find more tools for fixed cell imaging ›

The Image-iT Fixation/Permeabilization Kit contains:
• Fixative: high purity 4% formaldehyde (methanol-free) in PBS, pH 7.3
• Permeabilization solution: 0.5% Triton X-100
• Blocking buffer: 3% BSA, fraction V, de-lipidated, New Zealand source, in DPBS
• Wash solution: DPBS, no calcium, no magnesium

Specifications
Quantity1 Kit
Shipping ConditionAmbient
Product LineImage-iT
Unit SizeEach
Contents & Storage
• Image-iT Fixative solution
• Permeabilization solution
• Blocking solution
• Wash solution DPBS

Store at room temperature.