PCR Plate, 384-well, raised chimney
Thermo Scientific™

PCR Plate, 384-well, raised chimney

Thermo Scientific 384-well PCR Plate, with Raised Chimney—The Thermo Scientific Extra Volume 384-well full-skirted plate permits enables post-PCR applications suchRead more
Have Questions?
Catalog NumberQuantity
AB0937100 plates
Catalog number AB0937
Price (USD)
990.00
Each
Quantity:
100 plates
Request bulk or custom format
Price (USD)
990.00
Each
Thermo Scientific 384-well PCR Plate, with Raised Chimney—The Thermo Scientific Extra Volume 384-well full-skirted plate permits enables post-PCR applications such as enzymatic digestions and ethanol precipitation to be carried out in the wells due to the increased well volume.

Features of the Thermo Scientific 384-well PCR Plate, with Raised Chimney:

• Tall Chimney well design
• Increased well volume accommodates sequencing and wash steps
• Working well volume: 40 μL
• Cut corner: A24

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
PCR Plate, 384-well, raised chimney
PCR Plate, 384-well, raised chimney, barcoded, white
PCR Plate, 384-well, standard
PCR Plate, 384-well, standard, green
PCR Plate, 384-well, standard, purple
PCR Plate, 384-well, standard, white
PCR Plate, 384-well, standard, yellow
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging100 Plate
Product TypePCR Microplate
Quantity100 plates
Reaction SpeedStandard
Skirt StyleSkirted
Volume (Metric) Well40 μL
Volume (Metric) Working25 μL
BarcodeNo Barcode
ColorClear
MaterialPolypropylene
No. of Wells384
Unit SizeEach

Frequently asked questions (FAQs)

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What 96-well plates do you recommend for -80 degrees C storage?

Both polypropylene and polystyrene plates can be used to store samples at -80 degrees C and depends on the downstream application. Please note that the sealing method selected is critical for storing at these temperatures and heat sealing is recommended.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.