ABsolute Blue QPCR Mix, ROX
Thermo Scientific™

ABsolute Blue QPCR Mix, ROX

**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’veRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AB4138B
also known as AB-4138/B
1600 Reactions
AB4139A
also known as AB-4139/A
400 Reactions
Catalog number AB4138B
also known as AB-4138/B
Price (USD)
3,655.00
Each
No. of Reactions:
1600 Reactions
Price (USD)
3,655.00
Each
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of ABsolute Blue QPCR Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute Blue qPCR Mixes are 2X master mixes for probe chemistry. There are suitable alternatives available for all common real-time instruments. ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity. The master mixes incorporate an inert blue dye to enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

Highlights

• High performance—ABsolute sensitivity, consistency & convenience
• Enhanced contrast and reduced errors—easily identify correctly aliquotted reaction mix in all plates and tubes
• Universal—compatible with all major qPCR platforms
• Adaptable—available with different ROX passive reference dye concentrations

Applications

• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute Blue QPCR Mix, low ROX
ABsolute Blue QPCR Mix, with separate ROX vial
ABsolute Blue QPCR Mix, ROX
ABsolute Blue QPCR Mix, SYBR Green, low ROX
ABsolute Blue QPCR Mix, SYBR Green, ROX
ABsolute Blue QPCR Mix, SYBR Green, with separate ROX vial
ABsolute Blue QPCR Mix, SYBR Green, fluorescein
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
FormatTube
No. of Reactions1600 Reactions
PolymeraseTaq DNA Polymerase
Product LineABsolute Blue
Product TypeqPCR Mix
Quantity1,600 reactions
Sample TypeDNA, dsDNA
Sufficient For1600 Reactions
Volume20 mL
Concentration2X
Detection MethodPrimer-probe
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains:
• 16 x 1.25 mL ABsolute Blue QPCR ROX Mix (2X)
Sufficient for 1600 x 25 μL reactions.

Store at -20°C