Celleste™ 3D Analysis Module, network license (includes one seat)
Celleste™ 3D Analysis Module, network license (includes one seat)
Invitrogen™
Celleste™ 3D Analysis Module, network license (includes one seat)
This Celleste 3D Analysis Module, network license (includes one seat), helps visualize complex 3D objects using guided segmentation designed toRead more
Have Questions?
Catalog NumberQuantity
AMEP49181 each
Catalog number AMEP4918
Price (USD)
-
Request A Quote
Quantity:
1 each
This Celleste 3D Analysis Module, network license (includes one seat), helps visualize complex 3D objects using guided segmentation designed to facilitate identification and measurements. This module will enable users to threshold and generate iso-surfaces as well as measure distances, volumes, and areas, and perform other measurements. This module is used in conjunction with Celleste 6.0 Image Analysis Software.
Celleste 6 software is a suite of image analysis and visualization tools designed for a range of biological applications that lets you transform your images into quantitative data in a streamlined and customizable workflow. Coupled with the powerful image acquisition capabilities of our EVOS microscopes, Celleste 6 software allows you to seamlessly capture, process, measure, analyze, and share images and data. The 3D analysis module requires the use of the Celleste 3D Visualization Module, network license (includes one seat).
Celleste 6.0 software with the 3D Analysis Module offers you these important advantages:
• Icon-based wizard-driven workflow streamlines image analysis and eliminates guesswork• Preconfigured and optimized analysis templates for common applications help derive the most relevant results from image sets
• Wide range of report formats to visualize results, create reports, and share images and data
• Automatic segmentation of objects using any 2D frame in the Z-stack
• Seamless propagation t in Z with automated or semi-automated tools
• Automatic generation of iso-surfaces and pseudo-surfaces
• 3D counting, including co-localization
• Measurement of point-to-point distances, surfaces, and volumes
Celleste 6 software is a powerful image analysis solution that helps make your imaging research job easier and increases your productivity by seamlessly providing images and accurate data. Coupled with the powerful image acquisition capabilities of the EVOS imaging system, you have the tools that make it easy to capture, process, measure, analyze, and share your important images and data more effectively than ever.
Specifications
For Use With (Equipment)EVOS M5000, EVOSM7000 and Other Imaging Systems
Quantity1 each
Product LineCelleste
Unit SizeEach