EVOS™ Light Cube, Texas Red 2.0 - FAQs
EVOS™ Light Cube, Texas Red 2.0 - FAQs
View additional product information for EVOS™ Light Cube, Texas Red 2.0 - FAQs (AMEP4955)
4 product FAQs found
Which staining dyes can I use with my Countess 3 FL Automated Cell Counter?
EVOS light cubes, which enable fluorescent detection in the Countess 3 FL Automated Cell Counter, are compatible with a broad range of fluorescent dyes. Refer to our selection guide for an extensive list of compatible dyes. We also offer the ready-to-use ReadyCount Stains for easy viability staining.
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
How can I count different types of cells, such as yeast cells and PBMCs, with my Countess 3 Automated Cell Counter?
Countess 3 Automated Cell Counters produce accurate and precise counts across a range of cell types, including human and murine peripheral blood mononuclear cells (PBMCs). Cell types and isolated nuclei smaller than 5 microns in diameter may be counted in FL only mode. Fluorescent staining, compatible EVOS light cubes, and update to the latest software version is required.
For more information on EVOS Light Cubes and compatible dyes, please use the following link:
EVOS Cell Imaging System Accessories
Find additional tips, troubleshooting help, and resources within our Cell Counting Support Center.
Are EVOS 2.0 Light Cubes compatible with the Countess II FL?
Yes. All our EVOS Light Cubes can be used with the Countess II FL.
Are EVOS 2.0 Light Cubes compatible with older EVOS instruments?
Yes. Our EVOS 2.0 Light cubes can be used in all EVOS instruments with EVOS light cube functionality.