Search
| Catalog Number | Quantity |
|---|---|
| B13233 | 1 mg |
Penicillin-binding proteins, present on the cytoplasmic membranes of eubacteria, can be detected with BOCILLIN FL penicillin and BOCILLIN 650/665 penicillin.
Dissolve in high-quality anhydrous DMSO or water for a stock concentration of 1 to 10 mM.
For Research Use Only. Not for use in diagnostic procedures.