Experience nuclei counting with fluorescent-mode counting algorithms, simplified workflows, built-in dilution calculators and reports using the latest version of Countess software with optional 21 CFR Part 11 compliance. Download now!
Countess™ Test Beads
Invitrogen™

Countess™ Test Beads

Ensure accurate performance of the Countess Automated Cell Counter with Countess Test Beads. These latex microspheres have a nominal diameter of 10 μm ±10% and are supplied as a suspension containing 1 x 106 beads/mL ±10% to help verify that cell counting and sizing are within expected ranges.
Have Questions?
Catalog NumberQuantity
C102841 mL
Catalog number C10284
Price (USD)
156.00
Each
Quantity:
1 mL
Price (USD)
156.00
Each

The Countess Test Beads are 10 μm latex microspheres designed for use with the Countess Automated Cell Counter to verify accurate cell counting and sizing. The beads have a nominal diameter of 10 μm ±10% and are supplied as a suspension containing 1 x 106 beads/mL ±10%. To verify proper functioning of the Countess Automated Cell Counter, the beads are mixed 1:1 with trypan blue and loaded into the Countess Chamber Slide. The Countess Test Beads offer researchers a reliable tool to ensure their Countess Automated Cell Counter is performing proper cell counting and sizing analysis before running critical experiments.

We also offer the Countess 3 Standard Slide (Cat. No. A51876), which is a ready-to-use and reusable reference and demo slide for cell counting, viability, and fluorescence on the Countess Automated Cell Counters.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Detection MethodColorimetric
For Use With (Equipment)Countess Automated Cell Counter
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Label or DyeTrypan Blue
Quantity1 mL
Shipping ConditionRoom Temperature
Product LineCountess
TypeCountess Test Bead
Unit SizeEach
Contents & Storage
Contents: One ml of test beads at 1 x 106 beads/ml.

Storage: Store at 4 °C.