CellLight™ Late Endosomes-GFP, BacMam 2.0
Invitrogen™

CellLight™ Late Endosomes-GFP, BacMam 2.0

CellLight Late Endosomes-GFP, BacMam 2.0, provides an easy way to label late endosomes with green fluorescent protein (GFP) in liveRead more
Have Questions?
Catalog NumberQuantity
C105881 mL
Catalog number C10588
Price (USD)
710.00
Each
Quantity:
1 mL
Price (USD)
710.00
Each
CellLight Late Endosomes-GFP, BacMam 2.0, provides an easy way to label late endosomes with green fluorescent protein (GFP) in live cells. Simply add the reagent to your cells, incubate overnight, and the cells are ready to image in the morning.

Want to label other cell structures? Learn more about CellLight fluorescent protein labeling tools

This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to Rab7a. You can observe late endosomes-GFP behavior in live cells independently of organelle pH and label with multiple tracking or tracing dyes to image dynamic cellular processes.

Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

CellLight Technology is:
• Fast and convenient: simply add CellLight reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
• Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
• Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
• Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling

BacMam Technology

CellLight Late Endosomes-GFP, BacMam 2.0, is a fusion construct of Rab7a and emGFP, providing accurate and specific targeting to cellular Late Endosomes-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

Specifications
ColorGreen
Detection MethodFluorescence
Dye TypeGFP (EmGFP)
EmissionVisible
Excitation Wavelength Range488⁄510
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Fluorescent Imager
FormLiquid
Product LineCellLight, Molecular Probes
Quantity1 mL
Shipping ConditionWet Ice
Target FunctionEndocytosis
Label TypeFluorescent Dye
Product TypeEndosomes Label
Sub Cellular LocalizationEndosomes
Unit SizeEach
Contents & Storage
2°C to 6°C, protect from light. Do not freeze.

Frequently asked questions (FAQs)

How can I increase the transduction efficiency with the BacMam 2.0 reagents such as the the CellLight and Premo products?

Try varying particle-to-cell ratio (PPC), incubation volume, temperature and, cell density (if adherent cells are transduced). For adherent cells, we recommend a confluence of about 70%. Following the PPC, adjusting the volume is the next best parameter to change to optimize protein expression. If that doesn't work, you can also use the BacMam Enhancer Kit (Cat. No. B10107).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Is there any way to preserve the CellLights labeling beyond 5 days?

Cells transduced with the CellLights reagents can be stored frozen for several months after transduction, without loss of expression.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Are the CellLights products toxic to cells?

If the viral particles are used at the level we recommend, they are very well tolerated by cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

For how long will the CellLights products label my cells?

The BacMam 2.0 CellLights typically express for 5 days after transduction.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What cell types can the CellLights products be used with?

The first generation BacMam reagents were shown to efficiently transduce over 90 cell types, including stable cell lines and primary cells. With BacMam 2.0, it is now possible to also efficiently transduce primary neurons and stem cells.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (21)

Citations & References
Abstract
A Rab11a-enriched subapical membrane compartment regulates a cytoskeleton-dependent transcytotic pathway in secretory epithelial cells of the lacrimal gland.
Authors:Xu S, Edman M, Kothawala MS, Sun G, Chiang L, Mircheff A, Zhu L, Okamoto C, Hamm-Alvarez S,
Journal:J Cell Sci
PubMed ID:21984810
'Despite observations that the lacrimal gland has been identified as the principal source of dimeric immunoglobulin A (dIgA) in tears, the mechanism used by lacrimal gland acinar cells (LGACs) to transcytose dIgA produced by interstitial plasma cells is not well-characterized. This study identifies a transcytotic pathway in LGACs regulated by ... More
Visualizing the endocytosis of phenylephrine in living cells by quantum dot-based tracking.
Authors:Ma J, Wu L, Hou Z, Song Y, Wang L, Jiang W,
Journal:
PubMed ID:24855959
'To study the intracellular receptor-drug transportation, a fluorescent probe consisting of phenylephrine-polyethylene glycol-quantum dots conjugate was employed to track endocytosis process of phenylephrine in living cells. This type of movement was studied by continuously filming fluorescent images in the same cell. We also calculated the movement parameters, and divided the ... More
Particles on the move: intracellular trafficking and asymmetric mitotic partitioning of nanoporous polymer particles.
Authors:Yan Y, Lai ZW, Goode RJ, Cui J, Bacic T, Kamphuis MM, Nice EC, Caruso F,
Journal:
PubMed ID:23713907
'Nanoporous polymer particles (NPPs) prepared by mesoporous silica templating show promise as a new class of versatile drug/gene delivery vehicles owning to their high payload capacity, functionality, and responsiveness. Understanding the cellular dynamics of such particles, including uptake, intracellular trafficking, and distribution, is an important requirement for their development as ... More
Intracellular dynamics and fate of polystyrene nanoparticles in A549 Lung epithelial cells monitored by image (cross-) correlation spectroscopy and single particle tracking.
Authors:Deville S, Penjweini R, Smisdom N, Notelaers K, Nelissen I, Hooyberghs J, Ameloot M,
Journal:
PubMed ID:26164626
Novel insights in nanoparticle (NP) uptake routes of cells, their intracellular trafficking and subcellular targeting can be obtained through the investigation of their temporal and spatial behavior. In this work, we present the application of image (cross-) correlation spectroscopy (IC(C)S) and single particle tracking (SPT) to monitor the intracellular dynamics ... More
Probing site-exclusive binding of aqueous QDs and their organelle-dependent dynamics in live cells by single molecule spectroscopy.
Authors:Dong C, Chowdhury B, Irudayaraj J,
Journal:Analyst
PubMed ID:23493749
Understanding the biophysical and chemical interactions of nanoprobes and their fate upon entering live cells is critical for developing fundamental insights related to intracellular diagnostics, drug delivery and targeting. In this article we report herein a single molecule analysis procedure to quantitate site-specific exclusive membrane binding of N-acetyl-L-cysteine (NAC)-capped cadmium ... More