BODIPY&trade; 505/515 (4,4-Difluoro-1,3,5,7-Tetramethyl-4-Bora-3a,4a-Diaza-<i>s</i>-Indacene)
Invitrogen™

BODIPY™ 505/515 (4,4-Difluoro-1,3,5,7-Tetramethyl-4-Bora-3a,4a-Diaza-s-Indacene)

With its nonpolar structure and long-wavelength absorption and fluorescence, BODIPY™ 505/515 can be used as a stain for netural lipids and as a tracer for oil and other nonpolar lipids.
Have Questions?
Catalog NumberQuantity
D392110 mg
Catalog number D3921
Price (USD)
264.86
Special offer
Online exclusive
Ends: 31-Dec-2026
323.00
Save 58.14
Each
Quantity:
10 mg
Price (USD)
264.86
Special offer
Online exclusive
Ends: 31-Dec-2026
323.00
Save 58.14
Each

With its nonpolar structure and long-wavelength absorption and fluorescence, BODIPY™ 505/515 can be used as a stain for netural lipids and as a tracer for oil and other nonpolar lipids.

To make a concentrated stock solution from 1–5 mM, dissolve in either high quality anhydrous DMSO or 100% ethanol. Store frozen, desiccated and protected from light.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Excitation/Emission502/510 nm
Label TypeBODIPY Dyes
Molecular FormulaC13H15BF2N2
Molecular Weight (g/mol)248.08
Product LineBODIPY
Quantity10 mg
Recommended StorageStore in freezer (-5°C to -30°C) and protect from light.
Shipping ConditionRoom Temperature
Physical FormSolid
Product TypeFatty Acid
Unit SizeEach

Citations & References (27)

Citations & References
Abstract
A cluster of noninvoluting endocytic cells at the margin of the zebrafish blastoderm marks the site of embryonic shield formation.
Authors:Cooper MS,D'Amico LA
Journal:Developmental biology
PubMed ID:8948584
Flow cytometry-based binding assay for GPR40 (FFAR1; free fatty acid receptor 1).
Authors:Hara T, Hirasawa A, Sun Q, Koshimizu TA, Itsubo C, Sadakane K, Awaji T, Tsujimoto G,
Journal:Mol Pharmacol
PubMed ID:18927207
'GPR40 is a G protein-coupled receptor (GPCR) whose endogenous ligands have recently been identified as medium- and long-chain free fatty acids (FFAs), and it is thought to play an important role in insulin release. Despite recent research efforts, much still remains unclear in our understanding of its pharmacology, mainly because ... More
Fluorescent membrane probes incorporating dipyrrometheneboron difluoride fluorophores.
Authors:Johnson ID, Kang HC, Haugland RP
Journal:Anal Biochem
PubMed ID:1799206
'The spectroscopic properties of a new series of fatty acid analogs in which a dipyrrometheneboron difluoride fluorophore forms a segment of the acyl methylene chain are presented and their characteristics as fluorescent membrane probes are examined. When incorporated as a low mole fraction component in model phospholipid membranes, the probes ... More
Enhancement of BODIPY505/515 lipid fluorescence method for applications in biofuel-directed microalgae production.
Authors:Brennan L, Blanco Fernández A, Mostaert AS, Owende P,
Journal:J Microbiol Methods
PubMed ID:22521923
'This paper describes a microalgal cell lipid fluorescence enhancement method using BODIPY(505/515), which can be used to screen for lipids in wild-type microalgae and to monitor lipid content within microalgae production processes to determine optimal harvesting time. The study was based on four microalgae species (Dunaliella teteriolecta, Tetraselmis suecica, Nannochloropsis ... More
Determination of the depth of BODIPY probes in model membranes by parallax analysis of fluorescence quenching.
Authors:Kaiser RD, London E
Journal:Biochim Biophys Acta
PubMed ID:9767081
'The location of a series of lipophilic and lipid-attached BODIPY (4, 4-difluoro-4-bora-3a,4a-diaza-s-indacene) membrane probes was analyzed by the quenching of BODIPY fluorescence by a series of nitroxide-labeled lipids in which the depth of the nitroxide group is varied. When attached to the polar headgroup of PE the BODIPY remained near ... More