Fluo-4FF, AM, cell permeant - Special Packaging
Invitrogen™

Fluo-4FF, AM, cell permeant - Special Packaging

Fluo-4FF, AM, cell permeant - Special Packaging

Have Questions?
Catalog NumberQuantity
F2398110 x 50 μg
Catalog number F23981
Price (USD)
423.00
Each
Quantity:
10 x 50 μg
Request bulk or custom format
Price (USD)
423.00
Each

Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. Fluo-5F, fluo-5N, and fluo-4ff are analogs of fluo-4 with lower Ca2+-binding affinity, making them suitable for detecting intracellular calcium levels in the 1 μM to 1 mM range that would saturate the response of fluo-3 and fluo-4. Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells. These indicators are compatible with excitation at 488 nm by argon-ion laser sources, making them useful for confocal microscopy, flow cytometry, and microplate screening applications.

Calcium Indicator (AM Ester) Specifications:

  • Label (Ex/Em of Ca2+–bound form): Fluo-4ff (494/516 nm)
  • Fluorescence intensity increase upon binding Ca2+: >100 fold
  • Kd for Ca2+ in buffer: ∼9.7 μM
  • Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength

    Using TPEN to Control Heavy Metal Cations

    In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+. Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelatorTPEN (Cat. No. T1210).

    More Choices for Fluorescent Calcium Indicators

    We offer a large selection of Molecular Probes™ calcium indicators for use in various experimental scenarios. For more information, review Fluorescent Ca2+ Indicators Excited with Visible Light—Section 19.3 in the Molecular Probes™ Handbook.

    For UV-excitable Ca2+ indicators, protein-based Ca2+ indicators, conjugates of Ca2+ indicators, and for fluorescence-based indicators of other metal ions (i.e., Mg2+, Zn2+) review Indicators for Ca2+, Mg2+, Zn2+ and Other Metal Ions—Chapter 19 in the Molecular Probes™ Handbook.

  • For Research Use Only. Not for human or animal therapeutic or diagnostic use.
    Specifications
    Detection MethodFluorescence
    Dye TypeFluorescent Dye-Based
    Quantity10 x 50 μg
    Shipping ConditionRoom Temperature
    For Use With (Application)Cell Viability and Proliferation
    For Use With (Equipment)Confocal Microscope, Fluorescence Microscope, High Content Analysis Instrument, HTS Reader, Microplate Reader, Fluorescent Imager
    Product TypeDye
    Unit SizeEach
    Contents & Storage
    Store in freezer -5°C to -30°C and protect from light.

    Frequently asked questions (FAQs)

    I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

    The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

    After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

    Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

    Citations & References (17)

    Citations & References
    Abstract
    LIM domain only 4 (LMO4) regulates calcium-induced calcium release and synaptic plasticity in the hippocampus.
    Authors:Qin Z, Zhou X, Gomez-Smith M, Pandey NR, Lee KF, Lagace DC, Béïque JC, Chen HH,
    Journal:J Neurosci
    PubMed ID:22442089
    'The LIM domain only 4 (LMO4) transcription cofactor activates gene expression in neurons and regulates key aspects of network formation, but the mechanisms are poorly understood. Here, we show that LMO4 positively regulates ryanodine receptor type 2 (RyR2) expression, thereby suggesting that LMO4 regulates calcium-induced calcium release (CICR) in central ... More
    nNOS(+) striatal neurons, a subpopulation spared in Huntington's Disease, possess functional NMDA receptors but fail to generate mitochondrial ROS in response to an excitotoxic challenge.
    Authors:Canzoniero LM, Granzotto A, Turetsky DM, Choi DW, Dugan LL, Sensi SL,
    Journal:
    PubMed ID:23720635
    'Huntington''s disease (HD) is a neurodegenerative condition characterized by severe neuronal loss in the cortex and striatum that leads to motor and behavioral deficits. Excitotoxicity is thought to be involved in HD and several studies have indicated that NMDA receptor (NMDAR) overactivation can play a role in the selective neuronal ... More
    Mechanisms underlying cannabinoid inhibition of presynaptic Ca2+ influx at parallel fibre synapses of the rat cerebellum.
    Authors:Daniel H, Rancillac A, Crepel F
    Journal:J Physiol
    PubMed ID:15034129
    'Activation of CB1 cannabinoid receptors in the cerebellum acutely depresses excitatory synaptic transmission at parallel fibre-Purkinje cell synapses by decreasing the probability of glutamate release. This depression involves the activation of presynaptic 4-aminopyridine-sensitive K(+) channels by CB1 receptors, which in turn inhibits presynaptic Ca(2+) influx controlling glutamate release at these ... More
    Cross-tolerance to otherwise lethal N-methyl-D-aspartate and oxygen-glucose deprivation in preconditioned cortical cultures.
    Authors:Tauskela JS, Comas T, Hewitt K, Monette R, Paris J, Hogan M, Morley P
    Journal:Neuroscience
    PubMed ID:11720781
    'In vitro ischemic preconditioning induced by subjecting rat cortical cultures to nonlethal oxygen-glucose deprivation protects against a subsequent exposure to otherwise lethal oxygen-glucose deprivation. We provide evidence that attenuation of the postsynaptic N-methyl-D-aspartate (NMDA) receptor- and Ca(2+)-dependent neurotoxicity underlies oxygen-glucose deprivation tolerance. It is demonstrated that extended tolerance to otherwise ... More
    Mitochondrial calcium uniporter silencing potentiates caspase-independent cell death in MDA-MB-231 breast cancer cells.
    Authors:Curry MC, Peters AA, Kenny PA, Roberts-Thomson SJ, Monteith GR,
    Journal:Biochem Biophys Res Commun
    PubMed ID:23602897
    'The mitochondrial calcium uniporter (MCU) transports free ionic Ca(2+) into the mitochondrial matrix. We assessed MCU expression in clinical breast cancer samples using microarray analysis and the consequences of MCU silencing in a breast cancer cell line. Our results indicate that estrogen receptor negative and basal-like breast cancers are characterized ... More