Fura Red™, AM, cell permeant
Invitrogen™

Fura Red™, AM, cell permeant

Visible light-excitable fura-2 analog that offers particular possibilities for ratiometric measurement of Ca2+ in single cells
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
F302110 x 50 μg
F3020500 μg
Catalog number F3021
Price (USD)
671.65
Online exclusive
722.00
Save 50.35
Each
Quantity:
10 x 50 μg
Price (USD)
671.65
Online exclusive
722.00
Save 50.35
Each

Fura Red, AM is a visible light—excitable fura-2 analog that offers unique possibilities for ratiometric measurement of Ca2+ in single cells by microphotometry, imaging or flow cytometry when used with single excitation, green-fluorescent calcium indicators. This acetoxymethyl (AM) ester form is useful for noninvasive intracellular loading and is also available in 1 mg amounts (F-3020).

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Detection MethodFluorescence
Dye TypeFura Red
Emission Wavelength598 nm
Excitation Wavelength488 nm
Quantity10 x 50 μg
Shipping ConditionRoom Temperature
Emission Wavelength Range585 to 600 nm
Excitation Wavelength Range480 to 495 nm
For Use With (Application)Calcium Indicator
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineFura Red
Product TypeFura Red AM
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

When using Calcium Crimson, AM, as an indicator of intracellular calcium flux, I am not getting a good degree of change. The cells also have GFP. What can I do?

This is a known drawback of Calcium Crimson, AM (as well as Calcium Orange, AM and Fura Red, AM, which are also in the same emission range). You can try increasing the concentration and washing out of any unlabeled dye from the media, to try to get better signal-to-background. If that fails, we recommend using Rhod-3 AM instead, which has a much better change in signal in that wavelength.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Where can I find the manual for the Fura Red, AM, cell permeant (Cat. No. F3020, F3021)?

Unfortunately, we do not have a user manual for the Fura Red, AM, cell permeant (Cat. No. F3021). You can reference section 19.3 of the Molecular Probes handbook for recommendations for use.
There is also a large number of citations & references provided on the product page.

Basically, you can dissolve the product in good quality anhydrous DMSO to prepare a stock solution of 1-10 mM and use a final solution of about 1-10 µM in a physiological buffer. You can then incubate for 15 to 60 minutes, wash and then leave the cells for about 30 minutes to allow the AM group to hydrolyse before starting the assay. You can add Pluronic F-127 (e.g., Cat. No. P6866) to facilitate uptake. Store the DMSO stock solution at -20 degrees C.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (161)

Citations & References
Abstract
Mechanism of collagen activation in human platelets.
Authors:Roberts DE, McNicol A, Bose R
Journal:J Biol Chem
PubMed ID:14981087
The mechanism of collagen-induced human platelet activation was examined using Ca2+, Na+, and the pH-sensitive fluorescent dyes calcium green/fura red, sodium-binding benzofuran isophthalate, and 2',7'-bis(2-carboxyethyl)-5(6)-carboxyfluorescein. Administration of a moderate dose of collagen (10 microg/ml) to human platelets resulted in an increase in [Ca2+](i) and platelet aggregation. The majority of this ... More
Cluster formation of inositol 1,4,5-trisphosphate receptor requires its transition to open state.
Authors:Tateishi Y, Hattori M, Nakayama T, Iwai M, Bannai H, Nakamura T, Michikawa T, Inoue T, Mikoshiba K
Journal:J Biol Chem
PubMed ID:15583010
'The inositol 1,4,5-trisphosphate (IP(3)) receptor (IP(3)R) Ca(2+) channel plays pivotal roles in many aspects of physiological and pathological events. It was previously reported that IP(3)R forms clusters on the endoplasmic reticulum when cytosolic Ca(2+) concentration ([Ca(2+)](C)) is elevated. However, the molecular mechanism of IP(3)R clustering remains largely unknown, and thus ... More
Triad formation: organization and function of the sarcoplasmic reticulum calcium release channel and triadin in normal and dysgenic muscle in vitro.
Authors:Flucher BE, Andrews SB, Fleischer S, Marks AR, Caswell A, Powell JA
Journal:J Cell Biol
PubMed ID:8245124
'Excitation-contraction (E-C) coupling is thought to involve close interactions between the calcium release channel (ryanodine receptor; RyR) of the sarcoplasmic reticulum (SR) and the dihydropyridine receptor (DHPR) alpha 1 subunit in the T-tubule membrane. Triadin, a 95-kD protein isolated from heavy SR, binds both the RyR and DHPR and may ... More
Real time fluorescence imaging of PLC gamma translocation and its interaction with the epidermal growth factor receptor.
Authors:Matsuda M, Paterson HF, Rodriguez R, Fensome AC, Ellis MV, Swann K, Katan M
Journal:J Cell Biol
PubMed ID:11331309
'The translocation of fluorescently tagged PLC gamma and requirements for this process in cells stimulated with EGF were analyzed using real time fluorescence microscopy applied for the first time to monitor growth factor receptor--effector interactions. The translocation of PLC gamma to the plasma membrane required the functional Src homology 2 ... More
Vav1 and Ly-GDI two regulators of Rho GTPases, function cooperatively as signal transducers in T cell antigen receptor-induced pathways.
Authors:Groysman M, Hornstein I, Alcover A, Katzav S
Journal:J Biol Chem
PubMed ID:12386169
'The Rho family GTPases are pivotal for T cell signaling; however, the regulation of these proteins is not fully known. One well studied regulator of Rho GTPases is Vav1; a hematopoietic cell-specific guanine nucleotide exchange factor critical for signaling in T cells, including stimulation of the nuclear factor of activated ... More