Phalloidin Labeling Probes
Invitrogen™

Phalloidin Labeling Probes

Achieve precise and reliable F-actin staining with fluorescent and biotinylated phalloidins. Phalloidin conjugates are widely used in imaging applications to selectively label F-actin in a variety of sample types including fixed and permeabilized cells, tissue sections, and cell-free experiments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorExcitation Wavelength RangeDye Type
F432Green496/516 nmFITC (Fluorescein)
A22281Blue346/442 nmAlexa Fluor™ 350
A30104Violet405/450 nmAlexa Fluor™ Plus 405
A12379Green495/518 nmAlexa Fluor™ 488
O7466Green496/520 nmOregon Green™ 488
A22282Yellow531/554 nmAlexa Fluor™ 532
R415Red-orange540/565 nmTRITC
A22283Orange556/570 nmAlexa Fluor™ 546
A34055Orange555/565 nmAlexa Fluor™ 555
A30106Orange555/565 nmAlexa Fluor Plus 555
B3475Red558/569 nmBODIPY™
A12380Orange-red578/600 nmAlexa Fluor™ 568
A12381Red581/609 nmAlexa Fluor™ 594
T7471Red591/608 nmTexas Red™
A22284Far-red632/647 nmAlexa Fluor™ 633
A34054Far-red633/647 nmAlexa Fluor™ 635
A22287Far-red650/668 nmAlexa Fluor™ 647
A30107Far-red650/668 nmAlexa Fluor Plus 647
A22285Near-infrared663/690 nmAlexa Fluor™ 660
A22286Near-infrared679/702 nmAlexa Fluor™ 680
A30105Near-infrared758/784 nmAlexa Fluor™ Plus 750
B7474NoneNoneBiotin-XX
P3457NoneNonePhalloidin (unlabeled)
Catalog number F432
Price (USD)
583.65
Online exclusive
628.00
Save 44.35
Each
Color:
Green
Excitation Wavelength Range:
496/516 nm
Dye Type:
FITC (Fluorescein)
Price (USD)
583.65
Online exclusive
628.00
Save 44.35
Each
Fluorescent and biotinylated phalloidins are water soluble and bind to filamentous actin (F-actin) with nanomolar affinity, making them convenient probes for labeling, identifying, and quantifying F-actin in cryopreserved tissue sections, fixed and permeabilized cells, and cell-free experiments. Phalloidin conjugates bind similarly to actin from various species, including plants and animals, enabling staining of the cytoskeleton in a wide range of samples.

A variety of phalloidin conjugates for filamentous (F-actin) staining are available, including fluorescent Alexa Fluor and Alexa Fluor Plus phalloidins, along with phalloidins conjugated to classic fluorescent dyes such as BODIPY, fluorescein, and rhodamine. Phalloidin staining is spectrally compatible with other fluorescent stains used in cellular analyses such as GFP/RFP, Qdot nanocrystals, and other Alexa Fluor conjugates and antibodies. Biotin‐XX Phalloidin can be used to visualize actin filaments via fluorescent streptavidin tags or standard enzyme-mediated avidin/streptavidin techniques such as in electron microscopy. Unlabeled phalloidin is available for use as a control in blocking F‐actin staining or in promoting polymerization.

Phalloidin conjugates bind to both large and small actin filaments with similar affinity in a 1:1 stoichiometry between phallotoxin and actin subunits. They do not bind G-actin monomers.

Alexa Fluor and Alexa Fluor Plus phalloidin conjugates for F-actin staining

Fluorescent Alexa Fluor dye conjugates of phalloidin are popular F-actin stains, offering color choices across the full spectral range. These phalloidin conjugates provide researchers with fluorescent probes that are superior in brightness and photostability compared to other spectrally similar conjugates.

Alexa Fluor Plus Phalloidin conjugates retain the same specificity for actin but offer 3-5 times greater sensitivity and brightness compared to the corresponding Alexa Fluor Phalloidin conjugate. This increased brightness is beneficial for challenging F-actin imaging, such as the super‐resolution microscopy methods SIM and STORM, and for reliable staining of actin stress fibers.

Features of phalloidin probes

  • High specificity—binds selectively to F-actin, which allows for precise labeling of actin filaments in fixed cells and cryopreserved tissues
  • Strong affinity—nanomolar binding affinity for F-actin, which ensures stable and reliable actin staining
  • Extensive fluorescent conjugate options—over twenty conjugated varieties of phalloidin
  • Compatibility with fixed samples—typically used with fixed cells and tissues, making them suitable for actin staining in detailed structural studies, immunofluorescence staining, and IHC applications
  • Multiplexing capability—the wide availability of phalloidin conjugates enables their use in combination with other fluorescent probes and antibodies for multiplex imaging. Biotinylated phalloidin can be made use of in downstream streptavidin steps.
  • Quantitative analysis—can be used for quantitative analysis of F-actin distribution and density within cells, aiding in the study of cytoskeletal dynamics. The unlabeled phalloidin can be titrated as a control.
  • Ease of use—staining is straightforward and quick
  • Excellent stability—exhibit good photostability, which is essential for prolonged imaging sessions and time-lapse studies
  • Wide applicability—used for a range of applications, including studying cell morphology, motility, and the effects of drugs on the actin cytoskeleton
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Dye TypeFITC (Fluorescein)
Excitation Wavelength Range496/516 nm
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Confocal Microscope, Compatible with FITC/GFP filter set
Quantity300 Units
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Product TypePhalloidin
Sub Cellular LocalizationActin, Cytoskeleton
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?

One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you offer alternative package sizes for phalloidin or ReadyProbes reagents?

Alternative package sizes may be available through a custom request process. Contact Thermo Fisher Scientific customer support or your sales representative to discuss available options for the specific reagent and quantity needed.

Can I use conjugated phalloidin label probes on organoids?

Yes. Conjugated phalloidin labeling probes can be used with organoids when F-actin is accessible after fixation and permeabilization. Because organoids are thicker than monolayer cell cultures, protocol optimization may be needed, including permeabilization conditions, probe incubation time, and probe concentration.

Are conjugated phalloidin reagents antibodies?

No, Phalloidin is not an antibody. It is a bicyclic peptide toxin originally isolated from the Amanita phalloides mushroom and is used to bind and label F-actin in fixed and permeabilized samples.

Can I use methanol instead of DMSO to reconstitute the conjugated phalloidins from the lyophilized solids?

Yes, methanol can be used to reconstitute fluorescent phalloidin, but it creates a less concentrated stock solution than DMSO. Dissolve the vial contents in 1.5 mL of methanol to yield a 40X stock solution at a concentration of 200 assays/mL, which is equivalent to approximately 6.6 µM. One unit/assay of fluorescent phalloidins is equivalent to 5 µL of the methanolic stock solution. Methanol may be used as a stock-solution solvent, but methanol-containing fixatives are not recommended because they can disrupt F-actin.

Citations & References (459)

Citations & References
Abstract
The p42/p44 MAP kinase pathway prevents apoptosis induced by anchorage and serum removal.
Authors:Le Gall M,Chambard JC,Breittmayer JP,Grall D,Pouysségur J,Van Obberghen-Schilling E
Journal:Molecular biology of the cell
PubMed ID:10712523
Anchorage removal like growth factor removal induces apoptosis. In the present study we have characterized signaling pathways that can prevent this cell death using a highly growth factor– and anchorage-dependent line of lung fibroblasts (CCL39). After anchorage removal from exponentially growing cells, annexin V-FITC labeling can be detected after 8 ... More
Basilar papilla explants: a model to study hair cell regeneration-repair and protection.
Authors:Frenz DA, Yoo H, Liu W
Journal:Acta Otolaryngol
PubMed ID:9840500
Explants of basilar papillae from 6-7 days posthatch chicks were cultured in growth medium for a period of 1-8 days. Hair cells were counted following staining of stereocilia bundles with FITC-phalloidin, and the percentage of hair cell survival was determined by comparison to control (i.e. uncultured) specimens. Hair cell integrity ... More
The small GTP-binding protein Rac promotes the dissociation of gelsolin from actin filaments in neutrophils.
Authors:Arcaro A
Journal:J Biol Chem
PubMed ID:9422735
Gelsolin is an actin filament-capping protein that has been shown to play a key role in cell migration. Here we have studied the involvement of phosphoinositide 3-kinase (PI 3-kinase) and GTP-binding proteins (G-proteins) in the regulation of gelsolin-actin interactions in neutrophils. Inhibition of PI 3-kinase activity in vivo by wortmannin ... More
Integrin-mediated activation of focal adhesion kinase is independent of focal adhesion formation or integrin activation. Studies with activated and inhibitory beta3 cytoplasmic domain mutants.
Authors:Lyman S, Gilmore A, Burridge K, Gidwitz S, White GC
Journal:J Biol Chem
PubMed ID:9278407
Integrin alphaIIbbeta3 functions as the fibrinogen receptor on platelets and mediates platelet aggregation and clot retraction. Among the events that occur during either "inside-out" or "outside-in" signaling through alphaIIbbeta3 is the phosphorylation of focal adhesion kinase (pp125(FAK)) and the association of pp125(FAK) with cytoskeletal components. To examine the role of ... More
Responses induced by tacrine in neuronal and non-neuronal cell lines.
Authors:De Ferrari GV, von Bernhardi R, Calderón FH, Luza SC, Inestrosa NC
Journal:J Neurosci Res
PubMed ID:9589388
Alzheimer's disease (AD) is associated with a reduction in cholinergic activity as a result of specific neuronal loss. Current potential treatments for the disease include both cholinomimetic drugs and anticholinesterase inhibitors. One of the drugs approved by the FDA is tacrine (9-amine-1,2,3,4 tetrahydroacridine; THA), a strong acetylcholinesterase (AChE) inhibitor. We ... More