ImageDev Software Development Toolkit
ImageDev Software Development Toolkit
Thermo Scientific™
ImageDev Software Development Toolkit
ImageDev Software Toolkit is a highly optimized image processing library for the development of demanding 3D imaging applications such as medical & dental imaging, biology imaging, and industrial inspection and quality control. ImageDev provides software application developers an extensive library for processing, segmentation, and registration of scientific 2D and 3D images.
Have Questions?
Número de catálogo IMAGEDEV-SDK
Precio (CLP)
-
High-performance medical imaging development software applications, faster. Specifically designed for image-based scientific application developers, Thermo Scientific ImageDev software development toolkit (SDK) allows easy integration of advanced 2D and 3D+ image processing and analysis capabilities into imaging software applications – in medical, dental, life sciences, industrial inspection, materials science, and geosciences. ImageDev Toolkit provides software application developers with an extensive library for processing, segmentation, and registration of scientific 2D/3D+ images.
Find more information: ImageDev Software Development Toolkit
Key Features
Image analysis applications development
Accelerate your time-to-market: Easily integrate advanced image processing capabilities into your existing applications or into a new project, thanks to smart API design.
Build powerful image analysis workflows
Implement high-performance image processing workflows for large and complex data. Process large 2D and 3D images exceeding 32-bit addressing, such as “large-field” applications in microscopy.
Build image analysis foundations for the future
Think beyond tomorrow: Work with a true business partner for future-proof developments. Rely on strong support and innovation in image processing, image segmentation, and image registration in two, three, or more dimensions.
Integrate your analysis workflows with 2D/3D visualization
Take advantage of interoperability with any rendering engine. Utilize seamless integration with Thermo Scientific™ Open Inventor™ for full visualization and processing workflows.
Especificaciones
DescripciónHigh-performance medical imaging development software applications, faster.
TipoImaging Data Management Platform
Unit SizeEach