This product is animal origin and can only be shipped within the United States
Elastase, porcine pancreas aqueous suspension
Thermo Scientific Chemicals

Elastase, porcine pancreas aqueous suspension

Have Questions?
Catalog NumberQuantity
J61753.MC
also known as J61753-MC
100 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61753.MC
also known as J61753-MC
Price (USD)
-
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS39445-21-1
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofAu/mL Trypsin (BAEE): Report value
Assay from Supplier's CofAu/mL: Report value
CommentOne Unit cleaves one micromole of N-succinyl-L-alanyl-L-alanyl-L-alanine-p-nitroanilide per minute at 25°C, pH 8.0
FormAqueous suspension
Assay from Supplier's CofAEndotoxin EU/mL: Report value
View more
Elastase is used in combination with other proteases for protein analysis by mass spectrometry. Elastase, is used in tissue dissociation because elastin is found in highest concentrations in the elastic fibers of connective tissues, elastase is frequently used to dissociate tissues that contain extensive intercellular fiber networks. It is usually used with other enzymes such as collagenase, trypsin, and chymotrypsin, membrane protein solubilization, protein sequence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A serine protease that acts on elastin
  • Suitable for the isolation of Type II lung cells
RUO – Research Use Only
Contents & Storage
2°C to 8°C