Papain, Carica Papaya Latex
Papain, Carica Papaya Latex
Papain, Carica Papaya Latex
Thermo Scientific Chemicals
Papain, Carica Papaya Latex
A cysteine protease; breaks down intercellular matrix of cartilage | CAS: 9001-73-423 kd g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
1 g
Catalog number J61875.03
also known as J61875-03
Price (USD)
1,002.65
Online Exclusive
1,114.00
Save 111.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
1,002.65
Online Exclusive
1,114.00
Save 111.35 (10%)
Each
Chemical Identifiers
CAS9001-73-4
SpecificationsSpecification SheetSpecification Sheet
FormLyophilized
units/mgPActivates to at least 15.0 units per mg protein
Unit definitionOne unit hydrolyzes one micromole of benzoyl-L-arginine ethyl ester per minute at 25º C, pH 6.2, after activation in a solution containing 1.1 mM EDTA, 0.067 mM beta merceptoethanol and 5.5 mM cystein
Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
2°C to 8°C