This product is animal origin and can only be shipped within the United States
Chymotrypsinogen A, from bovine pancreas
Thermo Scientific Chemicals

Chymotrypsinogen A, from bovine pancreas

The zymogen of chymotrypsin | CAS: 9035-75-025.6 kd g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J64741.06
also known as J64741-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS9035-75-0
EINECS Number232-905-3
For Use With (Application)The zymogen of chymotrypsin
FormPowder
Molecular Weight (g/mol)25.6 kd
View more
Chymotrypsinogen A is used as a serine protease that hydrolyzes peptide bonds with aromatic or large hydrophobic side chains (Tyr, Trp, Phe, Met) on the carboxyl end of the peptide bond, used in the non-invasive determination of solid-state protein conformation using near infrared (NIR) spectroscopy. It has been used to study the partitioning of protein in polymer/polymer aqueous two-phase systems. The enzyme has also been used for self-interaction chromatography applications, to test the rapid measurement of protein osmotic second virial coefficients. In this technique, the protein is immobilized on chromatographic particles and its retention is measured using isocratic elution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
-30°C to -10°C