CloneJET PCR Cloning Kit
Thermo Scientific™

CloneJET PCR Cloning Kit

Thermo Scientific CloneJET PCR Cloning Kit, also available with DH10B cells (K123120), is an advanced positive selection system for high-efficiencyRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
K123240 Reactions
K123120 Reactions
Catalog number K1232
Price (USD)
339.65
Online exclusive
404.00
Save 64.35
Each
No. of Reactions:
40 Reactions
Request bulk or custom format
Price (USD)
339.65
Online exclusive
404.00
Save 64.35
Each
Thermo Scientific CloneJET PCR Cloning Kit, also available with DH10B cells (K123120), is an advanced positive selection system for high-efficiency cloning of PCR products generated with any thermostable DNA polymerase. Any other blunt or sticky-end DNA fragment can be cloned. It is ideal for phosphorylated or non-phosphorylated DNA fragments. Ligation into the included positive selection vector takes only 5 minutes, yielding more than 99% recombinant clones. Blunt-ended PCR products generated with a proofreading enzyme are ligated directly into the cloning vector.

PCR products generated either with non-proofreading DNA polymerases or mixtures of DNA polymerases are blunted prior to ligation in 5 minutes with the thermostable DNA Blunting Enzyme provided with the kit. All common laboratory E. coli strains can be directly transformed with the ligation product.

Features

The CloneJET PCR Cloning Kit contains a novel, ready-to-use positive selection cloning vector pJET1.2/blunt. The vector contains a lethal restriction enzyme gene that is disrupted by ligation of a DNA insert into the cloning site. As a result, only bacterial cells with recombinant plasmids are able to form colonies. Recircularized pJET1.2/blunt vector molecules lacking an insert express a lethal restriction enzyme, which kills the host E. coli cell after transformation. This positive selection drastically accelerates the process of colony screening and eliminates additional costs required for blue/white selection.

For convenience in mapping and manipulation of the insert, the pJET1.2/blunt cloning vector multiple cloning site contains two BglII recognition sequences that flank the insertion site. In addition, the vector contains a T7 promoter for in vitro and in vivo transcription as well as sequencing of the insert.

Highlights

• Fast—PCR cloning in only 5 minutes
• Highest efficiency – > 99% of positive clones
• No cloning background—positive selection vector
• Versatile—ideal for blunt-end or sticky-end cloning
• Economical—no expensive blue/white screening

Applications

• Cloning of blunt-end or 3'-dA tailed PCR products up to 10 kb
• Cloning of DNA fragments generated by restriction enzymes
• Sequencing of cloned DNA
• in vitro and in vivo transcription of cloned inserts from the T7 promoter

Includes

• pJET1.2/blunt Cloning Vector
• T4 DNA Ligase
• 2X Reaction Buffer
• DNA Blunting Enzyme
• pJET1.2 Forward Sequencing Primer (5'-CGACTCACTATAGGGAGAGCGGC-3')
• pJET1.2 Reverse Sequencing Primer (5'-AAGAACATCGATTTTCCATGGCAG-3')
• Control PCR Product
• Water, nuclease-free
• Detailed Protocol

Prior to electroporation, always column-purify the ligation mixture using e.g. GeneJET PCR Purification Kit #K0701 or chloroform to extract it. Electroporation is inhibited by the presence of proteins and salts in the mixture.

Related Products
CloneJET PCR Cloning Kit
Fast DNA End Repair Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialAmpicillin (AmpR)
Cloning MethodRestriction Enzyme⁄ MCS, Blunt PCR
For Use With (Application)PCR Cloning
No. of Reactions40 Reactions
Product LineCloneJET
Product TypeCloning Kit
Quantity40 Reactions
Sample TypePCR Products
VectorpJET1.2/blunt
FormatKit
Unit SizeEach

Frequently asked questions (FAQs)

Can I use the vector pJET1.2 for protein expression?

The pJET1.2 vector is not specifically designed for protein expression. It contains the T7 RNA promoter for in vitro transcription of the cloned insert. In some cases, this T7 promoter can be used for non-toxic gene expression in bacterial strains having IPTG inducible T7 RNA polymerase (like E. coli BL21); however, the promoter does not contain regulatory elements and protein expression will not be well regulated.

What are expected results of colony PCR when screening clones from the CloneJET kit?

Using pJET1.2 Forward and Reverse Sequencing Primers in PCR (as described in the protocol), negative colonies would yield PCR products that are about 120 bp long. Positive colonies would yield longer PCR products that should reflect the insert size (with addition of 120 bp from the vector).

Where can I expect the insert in the pJET1.2/blunt cloning vector after successful cloning?

The insert can be expected at the Eco32I (EcoRV) site in the vector at the position 369 since the pJET1.2/blunt cloning vector was linearized with Eco32I.

What type of DNA fragments can be cloned into the pJET1.2/blunt vector?

Any DNA fragment (either blunt- or sticky-end) that is 6 bp to 10 kb long and generated from PCR or restriction digestion, can be successfully cloned using the kit.

Do I need phosphorylated PCR Primers for CloneJET PCR Cloning Kit?

Phosphorylation of the PCR primers is not required since the 5'-ends of the pJET1.2/blunt vector contain phosphoryl groups for ligation.