Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD24 Antibodies

          Invitrogen

          CD24 Monoclonal Antibody (M1/69), NovaFluor™ Red 685, eBioscience™

          View all (120) CD24 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD24 Monoclonal Antibody (M1/69), NovaFluor™ Red 685, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          CD24 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD24 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD24 Antibody (M014T02R02-A) in Flow

          BALB/c mouse splenocytes were unstained (left) or stained with 0.8 µg of CD24 Monoclonal Antibody, NovaFluor Red 685 (right). All cells were co-stained with CD19 Monoclonal Antibody, eFluor 450 (Product # 48-0193-82). Total viable cells in the lymphocyte gate were used for analysis, as determined by LIVE/DEAD Blue (Product # L34962). Data was acquired on a 5-laser Cytek Aurora. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)
          CD24 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          M014T02R02-A

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.8 µg/test
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Rat / IgG2b, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          M1/69

          Conjugate

          NovaFluor™ Red 685 NovaFluor™ Red 685 NovaFluor™ Red 685
          • Unconjugated
          • Alexa Fluor 532 
          • Alexa Fluor 700 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 615
          • Brilliant UV 737
          • Brilliant UV 805
          • Brilliant Violet 711
          • Brilliant Violet 786
          • eFluor 450
          • FITC
          • Functional Grade
          • NovaFluor Blue 510
          • NovaFluor Blue 610-30S
          • NovaFluor Red 710
          • NovaFluor Yellow 570
          • NovaFluor Yellow 590
          • NovaFluor Yellow 660
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-Cyanine5.5
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Request custom conjugation

          Excitation/Emission Max

          637/685 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          25 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          RRID

          AB_3098990

          Product Specific Information

          Description
          The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow.

          This product contains 1 vial of NovaFluor conjugate and 1 vial of CellBlox Plus Blocking Buffer.

          Applications Tested
          The M1/69 antibody has been tested by flow cytometric analysis of mouse splenocytes and thymocytes. This can be used at less than or equal to 0.8 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Master mixes
          • Master mixes of NFs should be made at 2-8 °C and may be made up to 4 hours ahead of time.
          • We do not recommend storing master mixes containing NovaFluor conjugates overnight or longer.

          Whole Blood compatibility
          • When utilizing whole blood (as opposed to density-gradient-purified PBMC), we recommend lysing red blood cells in bulk prior to staining with NovaFluor conjugates.
          • See the Bulk Lysis of Human Whole Blood protocol here.
          • Staining of whole blood with NovaFluor conjugates followed by lysis of red blood cells may result in higher-than-expected background staining.

          Viability dye compatibility
          • NovaFluor dyes are not compatible with DNA intercalating viability dyes.
          • Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes.

          CellBlox Plus Blocking Buffer
          • This NovaFluor conjugate comes with CellBlox Plus Blocking Buffer (for 25 µg or 25 test products, Cat. No. C001T02F01; for 100 µg or 100 test, Cat. No. C001T03F01), essential for optimal staining.
          • Use CellBlox Plus Blocking Buffer in all experiments with NovaFluor conjugates.
          • Add 5 µL per sample to antibody cocktails/master mixes (regardless of how many Novafluor-conjugated antibodies are present) before combining with cells.
          • CellBlox Plus Blocking Buffer is compatible with either Super Bright Complete Blocking Buffer or Brilliant Stain Buffer and can be used in antibody cocktails/master mixes with those reagents.
          • For single-color controls, use 5 µL of CellBlox Plus Blocking Buffer per 100 µL of cell sample (10^3 to 10^8 cells).

          NovaFluor conjugates are based on Phiton technology utilizing novel fluorophore-containing nucleic acid dye structures that allow for engineered fluorescent signatures with consideration for spillover and spread impacts. Learn more

          Excitation: 637 nm; Emission: 685 nm; Laser: 633-640 nm (Red) Laser

          Target Information

          CD24 is a 35-50 kDa glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed on a variety of cell types, including erythrocytes, thymocytes, peripheral lymphocytes, and cells of the myeloid lineage. It is variably glycosylated, resulting in heterogeneity in molecular mass across different cell lineages, which can affect antibody staining levels on lymphocyte populations. In the context of B-cell development, CD24 is expressed from the pro-B-cell stage in the bone marrow through to mature, surface Ig-positive B cells, with very low or negative expression on plasma cells. It is also present on the majority of B-lineage acute lymphoblastic leukemias, B-cell chronic lymphocytic leukemias (CCLs), and B-cell non-Hodgkin's lymphomas. CD24 is thought to play a role in regulating B-cell proliferation and maturation, as well as in controlling autoimmunity. Additionally, CD24 has been reported to interact with P-selectin (CD62P), indicating a potential role in cell adhesion and signaling. CD24 is a valuable marker for studying B-cell development, leukemia, lymphoma, and autoimmune regulation, as well as its interactions with other molecules like P-selectin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cluster of differentiation 24; FLJ22950; FLJ43543; heat stable antigen; lymphocyte antigen 52; M1/69-J11D heat stable antigen; MGC75043; R13-Ag; unnamed protein product; X62 heat stable antigen

          View more View less

          Gene Aliases: Cd24; HSA; Ly-52; nectadrin

          View more View less

          UniProt ID: (Mouse) P24807

          View more View less

          Entrez Gene ID: (Mouse) 12484

          View more View less

          Function(s)
          protein binding protein kinase binding carbohydrate binding protein tyrosine kinase activator activity Hsp70 protein binding Hsp90 protein binding
          Process(es)
          response to hypoxia cell activation regulation of cytokine-mediated signaling pathway response to molecule of bacterial origin pre-B cell differentiation leukocyte migration involved in inflammatory response positive regulation of T cell mediated immune response to tumor cell positive regulation of inflammatory response to antigenic stimulus positive regulation of B cell apoptotic process cell adhesion heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules leukocyte cell-cell adhesion cell surface receptor signaling pathway positive regulation of cytosolic calcium ion concentration neuromuscular synaptic transmission negative regulation of neuroblast proliferation apoptotic mitochondrial changes positive regulation of neuron projection development cell migration regulation of cell-cell adhesion positive regulation of cell-cell adhesion apoptotic nuclear changes negative regulation of B cell proliferation neuron projection development T cell costimulation B cell receptor transport into membrane raft chemokine receptor transport out of membrane raft negative regulation of interleukin-6 production negative regulation of tumor necrosis factor production negative regulation of transforming growth factor beta3 production positive regulation of integrin activation positive regulation of cell adhesion mediated by integrin regulation of cell-cell adhesion mediated by integrin positive regulation of cell-cell adhesion mediated by integrin negative regulation of erythrocyte clearance homotypic cell-cell adhesion negative regulation of erythrocyte aggregation positive regulation of T cell homeostatic proliferation positive regulation of activated T cell proliferation cholesterol homeostasis regulation of MAPK cascade response to estrogen regulation of B cell differentiation negative regulation of neuron differentiation respiratory burst negative regulation of T cell homeostatic proliferation positive regulation of alpha-beta T cell proliferation synaptic vesicle endocytosis positive regulation of peptidyl-tyrosine phosphorylation negative regulation of neurogenesis positive regulation of calcium-mediated signaling negative regulation of monocyte chemotactic protein-1 production glomerular visceral epithelial cell differentiation glomerular parietal epithelial cell differentiation apoptotic signaling pathway negative regulation of inflammatory response to wounding positive regulation of inflammatory response to wounding positive regulation of nephron tubule epithelial cell differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-swbc5:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0