Measure-IT™ Thiol Assay Kit
Invitrogen™
Measure-IT™ Thiol Assay Kit
Provides easy and accurate quantitation of thiol
Have Questions?
Catalog NumberQuantity
M305501 Kit
Catalog number M30550
Price (USD)
693.65
Online exclusive
746.00
Save 52.35
Each
Quantity:
1 Kit
Price (USD)
693.65
Online exclusive
746.00
Save 52.35
Each
The Measure-iT™ Thiol Assay Kit proves easy and accurate quantitation of thiol. The assay has a linear range of 0.05-5 uM thiol, making it up to 400 times more sensitive than colorimetric methods based on Ellman's reagent. The assay is performed at room temperature; maximum fluorescence signal is attained in 5 minutes and is stable for at least 1 hour.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Thiol Quantitation
Product LineMeasure-iT
Quantity1 Kit
Shipping ConditionRoom Temperature
Product TypeThiol Assay Kit
Unit SizeEach
Citations & References (4)
Citations & References
Abstract
The ELISA Detectability and Potency of Pegfilgrastim Decrease in Physiological Conditions: Key Roles for Aggregation and Individual Variability.
Authors:
Journal:Sci Rep
PubMed ID:32051479
The choice of linker for conjugating R848 to inactivated influenza virus determines the stimulatory capacity for innate immune cells.
Authors:
Journal:Vaccine
PubMed ID:29398273
Natural Presentation of Glycosaminoglycans in Synthetic Matrices for 3D Angiogenesis Models.
Authors:
Journal:Front Cell Dev Biol
PubMed ID:34671601
Human CD64-targeted non-viral siRNA delivery system for blood monocyte gene modulation.
Authors:
Journal:Sci Rep
PubMed ID:28169353