Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Nestin Antibodies

          Invitrogen

          Nestin Monoclonal Antibody (10C2)

          Advanced Verification
          15 Published Figures
          56 References
          View all (59) Nestin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Nestin Monoclonal Antibody (10C2)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (19)
          • Published Figures (15)
          • Advanced Verification (3)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 37

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Nestin Antibody (MA1-110) in ICC/IF

          Immunofluorescent analysis of Nestin (green) in human neural stem cells derived from PD-3 iPSCs using Gibco® PSC Neural Induction Medium (Product # A1647801). The cells were fixed and permeabilized using Image-IT® Fixation/Permeabilization kit (Product # R37602), and blocked with the including blocking buffer for one hour at room temperature. Cells were stained with a Nestin monoclonal antibody (Product # MA1-110) at a dilution of 1:100 in blocking buffer for 3 hours at room temperature, and then incubated with a DyLight488-conjugated goat... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunohistochemistry (IHC)
          Nestin Antibody in Western Blot (WB)
          Nestin Antibody in Flow Cytometry (Flow)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Flow Cytometry (Flow)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunohistochemistry (IHC)
          Nestin Antibody in Immunohistochemistry (IHC)
          Nestin Antibody in Western Blot (WB)
          Nestin Antibody in Western Blot (WB)
          Nestin Antibody in Western Blot (WB)
          Nestin Antibody in Flow Cytometry (Flow)
          Nestin Antibody in Flow Cytometry (Flow)
          Nestin Antibody in Multiplex Immunohistochemistry (mIHC)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunocytochemistry (ICC/IF)
          Nestin Antibody in Immunohistochemistry (IHC)
          Nestin Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Nestin Antibody in Western Blot (WB)
          Nestin Antibody in Western Blot (WB)
          Nestin Antibody
          Nestin Antibody
          Nestin Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Nestin Monoclonal Antibody (10C2)

          Product Details

          MA1-110

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:4,000
          View 6 publications 6 publications

          Immunohistochemistry (IHC)

          -
          View 12 publications 12 publications

          Multiplex Immunohistochemistry (mIHC)

          1:20-1:100
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:100
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          Assay-dependent
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100
          View 33 publications 33 publications

          Flow Cytometry (Flow)

          1 µg/test
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          10C2

          Immunogen

          Human nestin protein corresponding to residues 1464-1614
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 555 
          • Alexa Fluor 647 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS with 1mg/mL BSA, 30% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2536821

          Product Specific Information

          MA1-110 has been successfully utilized in Western Blot, immunocytochemistry, flow cytometry, and immunofluorescence applications in human samples. MA1-110 also acts a neuronal stem cell marker and detects a band in Western blot at ~250 kDa.

          Target Information

          Nestin is a Class VI intermediate filament abundantly produced in the developing central nervous system (CNS) and in early embryonic neuroepithelial stem cells. Nestin has been widely used as a predominant marker for stem/progenitor cells, glioma cells, and tumor endothelial cells. Also, Nestin is a superior angiogenic marker to evaluate neovascularity of endothelial cells in tumors. Highly specific antibodies to human Nestin can aid in characterizing progenitor cells differentiating into distinct lineages, in enhancing the therapeutic potential of human neural stem/progenitor cells in the treatment of CNS diseases or injury, and in identifying neuroepithelial tumor cells. Nestin expression has been observed in pancreatic hematopoietic stem cell populations, and is considered a novel angiogenesis marker of proliferating endothelial cells in colorectal cancer tissue.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FLJ21841; intermediate filament protein; intermediate filament-associated protein RC2; unnamed protein product

          View more View less

          Gene Aliases: ESTM46; Ifaprc2; Marc2; Nbla00170; RC2

          View more View less

          UniProt ID: (Human) P48681, (Mouse) Q6P5H2

          View more View less

          Entrez Gene ID: (Human) 10763, (Mouse) 18008

          View more View less

          Function(s)
          protein binding intermediate filament binding CCR5 chemokine receptor binding
          Process(es)
          G2/M transition of mitotic cell cycle central nervous system development brain development positive regulation of intermediate filament depolymerization embryonic camera-type eye development negative regulation of protein binding negative regulation of catalytic activity stem cell proliferation positive regulation of neural precursor cell proliferation nervous system development negative regulation of neuron apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch