Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MHC Class II (I-Ak) Antibodies

          Invitrogen

          I-Ak Monoclonal Antibody (14V.18)

          2 References
          View all (4) MHC Class II (I-Ak) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite I-Ak Monoclonal Antibody (14V.18)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          I-Ak Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          I-Ak Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          I-Ak Antibody (MA1-70068) in Flow

          Flow cytometric analysis of spleen cells stained using a I-Ak monoclonal antibody (Product # MA1-70068) at a 0.5 µg/10^6 cells dilution. The isotype control was a biotin mouse IgG2a {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          I-Ak Antibody in Flow Cytometry (Flow)
          I-Ak Monoclonal Antibody (14V.18)

          Product Details

          MA1-70068

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          1 µg/1 million cells
          View 1 publication 1 publication

          Cytotoxicity Assay (Cytotoxicity)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          14V.18

          Immunogen

          A.TL.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • FITC
          • PE 
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          250 µg

          Purification

          Protein G

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1076835

          Product Specific Information

          MA1-70068 detects I-Ak in mouse samples.

          MA1-70068 has been successfully used in flow cytometry procedures.

          The MA1-70068 immunogen is A.TL.

          Store at 4°C. For long term storage, aliquot and freeze at -20°C, in volumes appropriate for single uses. Avoid freeze/thaw cycles.

          Target Information

          MHC Class II (I-Ak) serves as a critical component of the immune system, facilitating the recognition and response to extracellular pathogens and foreign substances. Its primary role is to present peptides derived from extracellular proteins to T cells for immune recognition. MHC Class II (I-Ak) is composed of an alpha and beta chain, both of which contribute to the peptide-binding groove. The location of MHC Class II (I-Ak) is restricted to antigen-presenting cells, such as dendritic cells, macrophages, and B cells. These cells are responsible for capturing and processing antigens from the extracellular environment. MHC Class II (I-Ak) consists of an extracellular domain, a transmembrane domain, and a cytoplasmic tail. The extracellular domain is responsible for interacting with the antigenic peptides, while the transmembrane domain anchors the MHC Class II molecule to the cell membrane. The cytoplasmic tail plays a role in intracellular signaling events.

          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: A alpha; H-2 class II histocompatibility antigen, A-B alpha chain; H-2 class II histocompatibility antigen, A-F alpha chain; H-2 class II histocompatibility antigen, A-K alpha chain; H-2 class II histocompatibility antigen, A-Q alpha chain; H-2 class II histocompatibility antigen, A-R alpha chain; H-2 class II histocompatibility antigen, A-S alpha chain; H-2 class II histocompatibility antigen, A-U alpha chain; histocompatibility 2, class II antigen A, alpha; IAalpha; major histocompatibility protein class II alpha chain; MHC class II antigen A alpha isoform-004; MHC-II I-A(d) alpha

          View more View less

          Gene Aliases: Aalpha; H-2Aa; H2-Aa; H2Aa; I-Aalpha; Ia-1; Ia1; IAalpha

          View more View less

          UniProt ID: (Mouse) P14434

          View more View less

          Entrez Gene ID: (Mouse) 14960

          View more View less

          Function(s)
          protein binding peptide antigen binding protein heterodimerization activity major histocompatibility complex protein
          Process(es)
          immune system process antigen processing and presentation of peptide or polysaccharide antigen via MHC class II immune response antigen processing and presentation antigen processing and presentation of exogenous peptide antigen via MHC class II response to interferon-gamma negative regulation of T cell proliferation positive regulation of T cell differentiation antigen processing and presentation of peptide antigen
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-794c4b9cd6-cc7sn:80/100.66.79.229:80.
          git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.52.0-Offline