Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD148 Antibodies

          Invitrogen

          CD148 Monoclonal Antibody (MEM-CD148/05), APC

          View all (15) CD148 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite CD148 Monoclonal Antibody (MEM-CD148/05), APC

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          CD148 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD148 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD148 Antibody (MA5-28590) in Flow

          Flow cytometry analysis (surface staining) of human peripheral blood using anti-CD148 (MEM-CD148/05) APC Monoclonal antibody (Product # MA5-28590). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD148 Antibody in Flow Cytometry (Flow)
          CD148 Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          CD148 Monoclonal Antibody (MEM-CD148/05), APC

          Product Details

          MA5-28590

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          10 µL/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          MEM-CD148/05

          Immunogen

          Human recombinant CD148 (amino acids 1-444)
          3D Epitope / Immunogen

          Conjugate

          APC APC APC
          • Unconjugated
          • Biotin 
          • PE 
          • Request custom conjugation

          Excitation/Emission Max

          651/660 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.07 mg/mL

          Purification

          Size-exclusion chromatography

          Storage buffer

          PBS, pH 7.4, with 0.2% BSA

          Contains

          15mM sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2745549

          Product Specific Information

          This antibody recognizes an extracellular epitope of CD148, a highly glycosylated up to 250 kDa receptor-like protein tyrosin phosphatase expressed mainly in lymphocytes, myeloid cells and epithelial cells.

          Recommended dilution is for 100 µL of whole blood or 1x10^6 cells in suspension.

          Target Information

          CD148 is a highly glycosylated transmembrane protein of up to 250 kDa, containing eight fibronectin type III domains in the extracellular region and one tyrosine phosphatase domain in the cytoplasmic region. Expression is found broadly on fibroblasts, epithelial cells, lymphocytes (B, T and NK cells), and myeloid cells (monocytes and granulocytes). CD148 can dephosphorylate proteins such as LAT and PLC-gamma, and is involved in the downregulation of T cell activation. Conversely, CD148 expression can be upregulated following T cell activation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD148 antigen; density-enhanced phosphatase 1; human density enhanced phosphatase-1; protein tyrosine phosphatase, receptor type, J polypeptide; protein-tyrosine phosphatase eta; susceptibility to colon cancer 1, mouse, homolog of

          View more View less

          Gene Aliases: CD148; DEP1; HPTP eta; HPTPeta; R-PTP-ETA; R-PTP-J; SCC1; THC10

          View more View less

          UniProt ID: (Human) Q12913

          View more View less

          Entrez Gene ID: (Human) 5795

          View more View less

          Function(s)
          protein tyrosine phosphatase activity platelet-derived growth factor receptor binding protein binding beta-catenin binding phosphatase activity protein kinase binding gamma-catenin binding cadherin binding mitogen-activated protein kinase binding delta-catenin binding
          Process(es)
          positive regulation of cell-matrix adhesion immune system process signal transduction axon guidance negative regulation of cell proliferation anatomical structure morphogenesis negative regulation of platelet-derived growth factor receptor signaling pathway cytokine-mediated signaling pathway hemopoiesis regulation of cell adhesion B cell differentiation platelet formation negative regulation of cell growth negative regulation of cell migration positive regulation of tumor necrosis factor production peptidyl-tyrosine dephosphorylation negative regulation of epidermal growth factor receptor signaling pathway negative regulation of vascular permeability negative regulation of MAP kinase activity positive regulation of MAPK cascade positive regulation of cell adhesion platelet-derived growth factor receptor signaling pathway positive regulation of phagocytosis positive regulation of immune response positive regulation of calcium-mediated signaling T cell receptor signaling pathway negative regulation of T cell receptor signaling pathway positive chemotaxis positive regulation of cellular component organization positive regulation of focal adhesion assembly positive regulation of protein kinase B signaling negative regulation of protein kinase B signaling contact inhibition positive regulation of Fc receptor mediated stimulatory signaling pathway positive regulation of intracellular signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-cbn8l:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline