Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SHANK1/SHANK3 Antibodies

          Invitrogen

          SHANK1/SHANK3 Monoclonal Antibody (N367/51), PerCP

          View all (15) SHANK1/SHANK3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SHANK1/SHANK3 Monoclonal Antibody (N367/51), PerCP

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          SHANK1/SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          SHANK1/SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SHANK1/SHANK3 Antibody (MA5-45675) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis using human neuroblastoma cells. Fixation involved 4% Formaldehyde for 15 min at RT. Samples were incubated with SHANK1/SHANK3 monoclonal antibody (Product # MA5-45675) at 1:100 for 60 min at RT, followed by Goat Anti-Mouse ATTO 488 at 1:100 for 60 min at RT. Counterstain used was Phalloidin Texas Red F-Actin stain; DAPI (blue) nuclear stain at 1:1,000; 1:5,00... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SHANK1/SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          SHANK1/SHANK3 Antibody in Immunocytochemistry (ICC/IF)
          SHANK1/SHANK3 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SHANK1/SHANK3 Monoclonal Antibody (N367/51), PerCP

          Product Details

          MA5-45675

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          N367/51

          Immunogen

          Fusion protein amino acids 538-626 (SH3 domain) of rat SHANK3.
          3D Epitope / Immunogen

          Conjugate

          PerCP PerCP PerCP
          • APC 
          • FITC
          • PE 
          • Request custom conjugation

          Excitation/Emission Max

          482/675 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          95.64mM phosphate/2.48mM MES, pH 7.4, with 0.5M EDTA

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2932129

          Product Specific Information

          Mouse: 100% identity (89/89 amino acids identical). Human: 97% identity (87/89 amino acids identical). ~70% identity with SHANK1 and SHANK2.

          1 µg/mL of MA5-45675 was sufficient for detection of SHANK1/SHANK3 in 20 µg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody.|Detects approximately 190kDa. Cross-reacts with SHANK1. Does not cross-react with SHANK2.

          This antibody was formerly sold as clone S367-51.

          Target Information

          Major scaffold postsynaptic density protein which interacts with multiple proteins and complexes to orchestrate the dendritic spine and synapse formation, maturation and maintenance. Interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and HOMER, respectively, and the actin-based cytoskeleton. Plays a role in the structural and functional organization of the dendritic spine and synaptic junction through the interaction with Arp2/3 and WAVE1 complex as well as the promotion of the F-actin clusters. By way of this control of actin dynamics, participates in the regulation of developing neurons growth cone motility and the NMDA receptor-signaling. Also modulates GRIA1 exocytosis and GRM5/MGLUR5 expression and signaling to control the AMPA and metabotropic glutamate receptor-mediated synaptic transmission and plasticity. May be required at an early stage of synapse formation and be inhibited by IGF1 to promote synapse maturation. {PubMed:21606927, PubMed:21795692, PubMed:23100419, PubMed:23897824, PubMed:24089484}.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: brain specific protein; brain-specific protein; GKAP/SAPAP-interacting protein; multidomain postsynaptic density protein; postsynaptic density protein; postsynaptic scaffold protein; proline rich synapse associated protein 2; proline-rich synapse-associated protein 2; SAPAP-interacting protein synamon; SH3/ankyrin domain gene 1; SH3/ankyrin domain gene 3; shank postsynaptic density protein; Shank postsynaptic density protein 3a; shank3 postsynaptic density protein; somatostatin receptor-interacting protein; SPANK-1; SPANK-2; proSAP2; proline-rich synapse-associated protein 2; SSTR-interacting protein; synamon

          View more View less

          Gene Aliases: DEL22q13.3; PROSAP2; PSAP2; SCZD15; Shank1a; SPANK-1; SPANK-2; Spank1; SSTRIP; synamon

          View more View less

          UniProt ID: (Human) Q9Y566, (Human) Q9BYB0, (Rat) Q9WV48, (Mouse) D3YZU1, (Rat) Q9JLU4, (Mouse) Q4ACU6

          View more View less

          Entrez Gene ID: (Human) 50944, (Human) 85358, (Rat) 78957, (Mouse) 243961, (Rat) 59312, (Mouse) 58234

          View more View less

          Function(s)
          G-protein coupled receptor binding protein binding SH3 domain binding receptor signaling complex scaffold activity GKAP/Homer scaffold activity somatostatin receptor binding ionotropic glutamate receptor binding identical protein binding macromolecular complex binding ankyrin repeat binding scaffold protein binding structural constituent of postsynaptic density zinc ion binding receptor binding
          Process(es)
          long-term memory associative learning adult behavior negative regulation of actin filament bundle assembly social behavior protein localization to synapse olfactory behavior habituation modulation of synaptic transmission synapse organization neuromuscular process controlling balance determination of affect righting reflex synapse maturation dendritic spine morphogenesis positive regulation of dendritic spine development protein-containing complex assembly vocalization behavior AMPA selective glutamate receptor signaling pathway maintenance of postsynaptic density structure positive regulation of excitatory postsynaptic potential MAPK cascade embryonic epithelial tube formation synapse assembly learning or memory learning memory locomotory behavior gene expression glial cell proliferation striatal medium spiny neuron differentiation exploration behavior locomotory exploration behavior locomotion vocal learning negative regulation of cell volume regulation of synaptic plasticity positive regulation of long-term neuronal synaptic plasticity brain morphogenesis sensory perception of touch positive regulation of synapse structural plasticity positive regulation of synaptic transmission, glutamatergic long-term synaptic potentiation long term synaptic depression regulation of dendritic spine morphogenesis neural precursor cell proliferation postsynaptic density assembly AMPA glutamate receptor clustering NMDA glutamate receptor clustering guanylate kinase-associated protein clustering response to interleukin-17 regulation of postsynapse organization regulation of long-term synaptic potentiation positive regulation of long-term synaptic potentiation positive regulation of glutamate receptor signaling pathway regulation of long term synaptic depression regulation of AMPA glutamate receptor clustering regulation of grooming behavior regulation of behavioral fear response cognition
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nbmw6:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline