Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Tau Antibodies

          Invitrogen

          Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8), Biotin

          Advanced Verification
          42 Published Figures
          410 References
          View all (227) Tau antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8), Biotin

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Published Figures (42)
          • Advanced Verification (1)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 43

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Tau (Ser202, Thr205) Antibody (MN1020B) in WB

          Molecular psychiatry 2024 - Abeta ice reduces Tau pathology. A Representative images of AT8-Thioflavine S double labeling showing AT8-positive neurites surrounding an amyloid plaque (Thioflavine). B Representative images of AT8 immunolabeling showing neuritic plaques in the brain of APP swe /PS1 dE9 mice after PBS, Abeta wt or Abeta ice inoculation in the dentate gyrus. C Representative images of Tau-positive neuritic plaques in the h... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8), Biotin

          Product Details

          MN1020B

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          -
          View 108 publications 108 publications

          Immunohistochemistry (IHC)

          -
          View 264 publications 264 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          -
          View 24 publications 24 publications

          ELISA (ELISA)

          Assay-dependent
          View 4 publications 4 publications

          GST Pull Down (GST-PD)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human

          Published species

          Cat, Chicken, Ferret, Fruit fly, Hamster, Horse, Human, Mammal, Mouse, Non-human primate, Rabbit, Rat, Rhesus monkey

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          AT8

          Immunogen

          Partially purified human PHF-Tau (aa 202, 205).
          3D Epitope / Immunogen

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_223648

          Product Specific Information

          MN1020B targets PHF-tau (Ser202/Thr205) in IF, ELISA, and IHC (P) applications and shows reactivity with Human samples.

          The MN1020B immunogen is partially purified human PHF-Tau.

          MN1020B detects PHF-tau (Ser202/Thr205)a which has a predicted molecular weight of approximately 79 kDa.

          Target Information

          Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: FLJ31424; FTDP17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; MGC138549; microtubule-associated protein tau; microtubules; neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHF-tau; PHFtau; protein phosphatase 1, regulatory subunit 103; Tau-4; Tau-derived paired helical filament hexapeptide; Tau5; unnamed protein product

          View more View less

          Gene Aliases: DDPAC; FTD1; FTDP-17; MAPTL; MSTD; MTBT1; MTBT2; PPND; PPP1R103; TAU; tau-40; Tau-PHF6

          View more View less

          UniProt ID: (Human) P10636

          View more View less

          Entrez Gene ID: (Human) 4137

          View more View less

          Function(s)
          DNA binding AT DNA binding double-stranded DNA binding single-stranded DNA binding RNA binding actin binding enzyme inhibitor activity protein binding microtubule binding tubulin binding SH3 domain binding enzyme binding protein kinase binding protein binding, bridging apolipoprotein binding dynactin binding phosphatidylinositol binding identical protein binding sequence-specific DNA binding chaperone binding protein phosphatase 2A binding Hsp90 protein binding lipoprotein particle binding histone-dependent DNA binding microtubule lateral binding microtubule stabilizing activity phosphatidylinositol bisphosphate binding
          Process(es)
          microtubule cytoskeleton organization microglial cell activation cellular response to DNA damage stimulus cell-cell signaling synapse assembly learning or memory memory response to lead ion regulation of autophagy negative regulation of gene expression negative regulation of mitochondrial membrane potential rRNA metabolic process axonal transport of mitochondrion central nervous system neuron development regulation of microtubule polymerization or depolymerization regulation of microtubule polymerization positive regulation of microtubule polymerization cytoplasmic microtubule organization neuron projection development positive regulation of superoxide anion generation regulation of chromosome organization stress granule assembly cellular response to heat cellular response to reactive oxygen species positive regulation of axon extension microtubule polymerization astrocyte activation regulation of synaptic plasticity intracellular distribution of mitochondria generation of neurons synapse organization regulation of calcium-mediated signaling protein polymerization protein homooligomerization regulation of microtubule-based movement axon development regulation of microtubule cytoskeleton organization plus-end-directed organelle transport along microtubule regulation of mitochondrial fission negative regulation of mitochondrial fission fibril organization axonal transport regulation of cellular response to heat regulation of long term synaptic depression positive regulation of protein localization to synapse neurofibrillary tangle assembly negative regulation of establishment of protein localization to mitochondrion positive regulation of cellular protein localization amyloid fibril formation cellular response to nerve growth factor stimulus cellular response to brain-derived neurotrophic factor stimulus
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-xbl4w:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0