Product Image
Thermo Scientific™

ELISA Wash Buffer (30X)

ELISA Wash Buffer is a Tris-based wash buffer for use in cytokine and other sandwich ELISA procedures. The buffer isRead more
Have Questions?
Catalog NumberQuantity
N50350 mL
Catalog number N503
Price (USD)
123.65
Online exclusive
128.00
Save 4.35
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
123.65
Online exclusive
128.00
Save 4.35
Each
ELISA Wash Buffer is a Tris-based wash buffer for use in cytokine and other sandwich ELISA procedures. The buffer is the same formulation that is supplied with most Pierce ELISA kits for cytokines and chemokines. The 30X concentrated stock solution is diluted 1:30 (vol:vol) in ultrapure water to yield sufficient wash buffer for a typical 96-well microplate ELISA protocol. Supplement an existing ELISA kit or use as a convenient general purpose wash buffer for customized ELISA methods.

Preparation: Thaw at room temperature (22–25°C). In a clean glass or plastic container, dilute the ELISA Wash Buffer concentrate 1:30 (vol:vol) with distilled or deionized water. Mix thoroughly. Equilibrate 1X wash buffer to room temperature before use in an assay. Prepare new 1X wash buffer for each day of use.

For a complete kit of ELISA buffers, see ELISA Buffer Kit.
View all available buffers and reagents for ELISA applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)ELISA
FormLiquid
Product TypeWash Buffer
Quantity50 mL
Unit SizeEach
Contents & Storage
Store at -20°C.