Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HMGA2 Antibodies

          Invitrogen

          HMGA2 Polyclonal Antibody

          Advanced Verification
          4 Published Figures
          3 References
          View all (4) HMGA2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite HMGA2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          • Published Figures (4)
          • Advanced Verification (1)
          HMGA2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          HMGA2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 17

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          HMGA2 Antibody (PA5-21320) in ICC/IF

          HMGA2 antibody detects HMGA2 protein at nucleus by immunofluorescent analysis. Sample: HeLa cells were fixed in 4% paraformaldehyde at RT for 15 min. Green: HMGA2 protein stained by HMGA2 antibody (Product # PA5-21320) diluted at 1:500. Red: Phalloidin, a cytoskeleton marker, diluted at 1:200. Scale bar = 10 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          HMGA2 Antibody in Immunocytochemistry (ICC/IF)
          HMGA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HMGA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HMGA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HMGA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HMGA2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody in Immunoprecipitation (IP)
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody in Immunohistochemistry (IHC)
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody in Western Blot (WB)
          HMGA2 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          PA5-21320

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -

          Immunoprecipitation (IP)

          1:100-1:500
          -

          ChIP assay (ChIP)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein encompassing a sequence within the center region of human HMGA2. The exact sequence is proprietary.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.13 mg/mL

          Amount

          13 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          RRID

          AB_11155799

          Product Specific Information

          Recommended positive controls: A431, HeLa, HepG2, HCT116.

          Predicted reactivity: Mouse (95%), Rat (92%), Zebrafish (85%), Xenopus laevis (80%), Chicken (98%).

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          HMGA2, previously known as HMGI-C, belongs to the group of high mobility chromosomal proteins that have the AT-hook DNA-binding motif. It is highly and ubiquitously expressed during embryogenesis. Chromosomal rearrangements of the HMGA2 gene and overexpression are frequently associated with benign and malignant tumors, respectively. Human and mouse HMGA2 shares 95% amino acid sequence identity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: high mobility group AT-hook protein 2; high mobility group protein I; HMGA2/KRT121P fusion; HMGI-C; non-histone chromosomal architectural protein HMGI-C; unnamed protein product

          View more View less

          Gene Aliases: 9430083A20Rik; BABL; HMGI-C; HMGIC; LIPO; pg; pygmy; SRS5; STQTL9

          View more View less

          UniProt ID: (Human) P52926, (Mouse) P52927

          View more View less

          Entrez Gene ID: (Human) 8091, (Rat) 84017, (Mouse) 15364

          View more View less

          Function(s)
          transcription regulatory region sequence-specific DNA binding nucleic acid binding DNA binding AT DNA binding transcription cofactor activity transcription corepressor activity DNA-(apurinic or apyrimidinic site) lyase activity protein binding DNA binding, bending enzyme binding nucleosomal DNA binding cAMP response element binding MH2 domain binding MH1 domain binding SMAD binding 5'-deoxyribose-5-phosphate lyase activity RNA polymerase II sequence-specific DNA binding transcription factor binding C2H2 zinc finger domain binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter epithelial to mesenchymal transition chondrocyte differentiation mesodermal-endodermal cell signaling base-excision repair regulation of transcription, DNA-templated spermatogenesis positive regulation of cell proliferation male gonad development response to virus regulation of gene expression regulation of cell cycle process positive regulation of gene expression cell proliferation in forebrain pituitary gland development negative regulation of Wnt signaling pathway adrenal gland development heterochromatin assembly negative regulation of intracellular steroid hormone receptor signaling pathway somatic stem cell population maintenance multicellular organism growth intracellular signal transduction endodermal cell differentiation chondrocyte proliferation positive regulation of multicellular organism growth negative regulation of apoptotic process negative regulation of DNA binding negative regulation by host of viral transcription fat cell differentiation positive regulation of angiogenesis negative regulation of single stranded viral RNA replication via double stranded DNA intermediate negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of JAK-STAT cascade mesodermal cell differentiation negative regulation of astrocyte differentiation mesenchymal cell differentiation stem cell differentiation regulation of growth hormone secretion lung epithelium development epithelial tube branching involved in lung morphogenesis positive regulation of epithelial cell proliferation involved in lung morphogenesis adipose tissue development fat pad development positive regulation of cell proliferation in bone marrow positive regulation of protein serine/threonine kinase activity regulation of peptide hormone secretion oncogene-induced cell senescence regulation of stem cell population maintenance positive regulation of stem cell proliferation negative regulation of cellular senescence negative regulation of double-strand break repair via nonhomologous end joining chromatin organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5799cd5976-pbfh7:80/100.66.77.38:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0