Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Ferroportin Antibodies

          Invitrogen

          Ferroportin Polyclonal Antibody

          6 Published Figures
          16 References
          View all (13) Ferroportin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Ferroportin Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          • Published Figures (6)
          Ferroportin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Ferroportin Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 13

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ferroportin Antibody (PA5-22993) in ICC/IF

          Immunocytochemistry analysis of Ferroportin in HepG2 cells. Samples were incubated with Ferroportin polyclonal antibody (Product # PA5-22993) using a dilution of 5.0 µg/mL overnight at 4°C followed by anti-rabbit DyLight 488 (green) at a dilution of 1:500. Cells were fixed and permeabilized for 10 minutes using -20°C MeOH. Alpha tubulin (DM1A) was used as a co-stain at a 1:1,000 dilution and detected with a... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ferroportin Antibody in Immunocytochemistry (ICC/IF)
          Ferroportin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ferroportin Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Ferroportin Antibody in Western Blot (WB)
          Ferroportin Antibody in Flow Cytometry (Flow)
          Ferroportin Antibody in Flow Cytometry (Flow)
          Ferroportin Antibody in Flow Cytometry (Flow)
          Ferroportin Antibody in Immunohistochemistry (IHC)
          Ferroportin Antibody in Western Blot (WB)
          Ferroportin Antibody in Western Blot (WB)
          Ferroportin Antibody in Western Blot (WB)
          Ferroportin Antibody in Western Blot (WB)
          Ferroportin Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Ferroportin Polyclonal Antibody

          Product Details

          PA5-22993

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          View 10 publications 10 publications

          Immunohistochemistry (IHC)

          -
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:200
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Pig, Rat

          Published species

          Human, Mouse, Rat, Rhesus monkey

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide made to an internal portion of human Ferroportin 1 (within residues 250-300).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11154326

          Product Specific Information

          Suggested positive control: human intestine protein.

          Target Information

          SLC40A1 is an iron-regulated transporter assumed to play roles in intestinal iron absorption and cellular iron release. It mediates iron efflux in the presence of ferroxidase. Defects in SLC40A1 cause hemochromatosis type 4 (HFE4), which is an autosomal dominant iron-loading disorder.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CAR1; cell adhesion regulator; duodenal-specific; ferroportin 1; ferroportin 1; ireg1; ferroportin-1; ferroportin1; FPN-N; iron regulated gene 1; iron-regulated transporter 1; metal transporter protein 1; metal transporting protein 1; polycythaemia; putative cellular iron exporter; SLC11A3 iron transporter; SLC40 iron transporter; SLC40A1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 3; solute carrier family 39 (iron-regulated transporter), member 1; solute carrier family 40 (iron-regulated transporter), member 1; solute carrier family 40 member 1; transporter; transporter; Fpn1; unnamed protein product

          View more View less

          Gene Aliases: Dusg; FPN; FPN1; HFE4; IREG1; MST079; MSTP079; MTP; MTP1; Ol5; Pcm; SLC11A3; Slc39a1

          View more View less

          UniProt ID: (Human) Q9NP59, (Mouse) Q9JHI9, (Rat) Q923U9

          View more View less

          Entrez Gene ID: (Bovine) 527023, (Pig) 100737517, (Human) 30061, (Mouse) 53945, (Rat) 170840

          View more View less

          Function(s)
          iron ion transmembrane transporter activity protein binding ferrous iron transmembrane transporter activity peptide hormone binding identical protein binding secondary carrier transporter
          Process(es)
          cellular iron ion homeostasis iron ion transmembrane transport transmembrane transport multicellular organismal iron ion homeostasis ferrous iron export lymphocyte homeostasis endothelium development iron ion transport negative regulation of apoptotic process positive regulation of transcription from RNA polymerase II promoter spleen development spleen trabecula formation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch