PowerEase™ Touch HV Power Supply, 115 VAC
PowerEase™ Touch HV Power Supply, 115 VAC
PowerEase™ Touch HV Power Supply, 115 VAC
Invitrogen™
PowerEase™ Touch HV Power Supply, 115 VAC
Invitrogen PowerEase Touch HV Power Supply is a high-voltage power supply that is operated using a touch-screen user interface. TheRead more
Have Questions?
Catalog NumberQuantity
PSHV001 each
Catalog number PSHV00
Price (USD)
4,013.65
Online exclusive
4,225.00
Save 211.35
Each
Quantity:
1 each
Price (USD)
4,013.65
Online exclusive
4,225.00
Save 211.35
Each
Invitrogen PowerEase Touch HV Power Supply is a high-voltage power supply that is operated using a touch-screen user interface. The 4.3-inch backlit LCD touchscreen and intuitive user interface design make setting up custom protocols or selecting one of the several pre-programmed gel electrophoresis and transfer methods a breeze. This power supply is ideal for high-voltage applications like 2D electrophoresis or isoelectric focusing. It also supports high-throughput DNA/RNA electrophoresis, SDS-PAGE, native PAGE, and western blotting.
Learn more about 2D electrophoresis ›
Learn more about available power supplies ›
The PowerEase Touch HV Power Supply offers four sets of output jacks that can be used simultaneously and four operation modes: constant voltage, voltage ramp, constant current, and constant power for flexibility of use and efficiency. Each high-voltage power supply comes with four PowerEase Touch HV Adapter sets that attach to the electrical leads of some electrophoresis equipment to enable compatibility, including the ZOOM IPGRunner Mini-Cell, XCell SureLock Mini-Cell, Mini Gel Tank, and the XCell4 SureLock Midi-Cell.
Features of the PowerEase Touch HV Power Supply include:
• LCD touchscreen display and user interface with clear menu prompts for easy use by hand or stylus
• Advanced UI/UX design combined with 4.3-inch backlit LCD touchscreen for ease of use and convenience in monitoring run progress
• Maximum output of 3500 V, 500 mA, and 250 W
• Four sets of output terminals for running of multiple electrophoresis units
• Four sets of the PowerEase Touch HV adapters for use with Invitrogen electrophoresis tanks
• Pop-up feet allowing change to the screen viewing angle
• Sturdy polyurethane feet and stackable housing design
• Running modes that include constant voltage, voltage ramp, constant current, or constant power
• Fully programmable, with either constant voltage, current, or power
• Flexibility to either program a custom method (up to 100 custom methods, 20 steps per method, 999 minutes per step) or select one of several pre-programmed gel electrophoresis and transfer methods
• User adjustable device settings such as beep volume, screen brightness, and sleep mode duration
• Automatic safety features including No Load, Over Temperature, Over Voltage, Over Current, Load Change, and Ground Leak detection
• Ability to turn off No Load Detection for low current applications like isoelectric focusing with IPG strips
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Electrical Requirements115 V
Max. Current500 mA
Quantity1 each
Shipping ConditionRoom Temperature
For Use With (Application)Gel Electrophoresis, IPG Strips, IEF gels
For Use With (Equipment)Zoom IPGRunner, Electrophoresis tanks
Max. Voltage3500 V
Product LinePowerEase
Unit SizeEach
Contents & Storage
• Power supply
• 4 sets of adaptors
• Quick reference card
Frequently asked questions (FAQs)
Do you offer a power supply that is comparable to the Bio-Rad PowerPac HV Power Supply?
Yes, we recommend using the PowerEase Touch HV Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Do you have an alternative for the discontinued ZOOM Dual Power Supply?
We offer the PowerEase Touch HV Power Supply as an alternative for the discontinued ZOOM Dual Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Do I need to use PowerEase Touch HV Power Supply Adapters with the PowerEase Touch HV Power Supply even if I have Invitrogen gel tanks?
The PowerEase Touch HV Power Supply Adapters are designed to facilitate the connection of some electrophoresis tanks to the PowerEase Touch HV Power Supply. The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply. The supported electrophoresis tanks include:
- ZOOM IPGRunner Mini-cell
- XCell SureLock Mini-Cell
- Mini Gel Tank
- XCell4 SureLock Midi-Cell
Note: The PowerEase Touch HV Power Supply Adapters are not needed for use of the SureLock Tandem Midi Gel Tank with the PowerEase Touch HV Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Why does the PowerEase Touch HV Power Supply come with power supply adapters?
For safety purposes, the output jacks of the PowerEase Touch HV Power Supply have been recessed compared to other PowerEase Touch power supplies. This requires a firm elongation of the leads on the cables of the tank and this is achieved using the power supply adapters that are provided. The adapters are designed to remain on the electrophoresis tank electrical leads when the lid is removed from the power supply.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
How many gels can I simultaneously transfer using the PowerEase Touch HV Power Supply?
The PowerEase Touch HV Power Supply can be used for simultaneous transfer of 2 mini gels. We do not recommend using it for transfer of midi gels.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.