Proteinase K (50 μg/μL)
Proteinase K (50 μg/μL)
Proteinase K (50 μg/μL)
Invitrogen™
Proteinase K (50 μg/μL)
This Proteinase K is used in QuantiGene assays and is one of two components in the QuantiGene Sample Processing kits.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
QS0511750 μL
QS0510150 μL
QS05123.5 mL
Catalog number QS0511
Price (USD)
355.65
Online exclusive
382.00
Save 26.35
Each
Quantity:
750 μL
Price (USD)
355.65
Online exclusive
382.00
Save 26.35
Each
This Proteinase K is used in QuantiGene assays and is one of two components in the QuantiGene Sample Processing kits. Proteinase K digests proteins such as ribonucleases that could interfere with the quantitation of RNA using QuantiGene assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Quantity750 μL
Shipping ConditionBlue Ice
Product TypeProteinase K
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).