Product Image
Thermo Scientific™
Phenom™ Quartz PCI/CFR Software
Phenom Quartz PCI/CFR Software, available on all Phenom Desktop SEMs including the Phenom Pro, ProX, XL G2, and Pharos Desktop SEMs, helps you meet the requirements of 21 CFR Part 11.
Have Questions?
Catalog number QUARTZ-PCI-CFR
Price (MXN)
-
Thermo Scientific™ Phenom™ Quartz PCI/CFR Software, available on all Phenom Desktop SEMs including the Thermo Scientific Phenom Pro, ProX, XL G2, and Pharos Desktop SEMs, helps you meet the requirements of 21 CFR Part 11. The integration of the Phenom Desktop SEM with the Quartz PCI/CFR Software allows direct acquisition of both navigational (optical) camera and SEM images. This significantly reduces the complexity of your standard operating procedures (SOPs), ensuring compliance with data integrity requirements.
After selecting the region of interest and the required imaging parameters in the Phenom user interface, you are able to acquire your images with the Quartz PCI/CFR Software.
Benefits​
  • Direct image acquisition from the Phenom Desktop SEM.
  • Various post-acquisition image processing tools like adjusting the brightness and contrast, local smoothing and sharpening tools, and annotation and drawing tools are available.
  • All changes that are applied to images are logged in an audit trail system.