Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%).
This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83334. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
As an L-hydroxy-specific enzyme, EHHADH (enoyl-CoA-hydratase:3-hydroxyacyl-CoA dehydrogenase), also known as Peroxisomal L-bifunctional enzyme, is a 723 amino acid protein has an essential tripeptide sequence on its carboxyl-terminus that is required for peroxisomal transport. EHHADH-null mice only exhibit a blunted peroxisome proliferative response when challenged with a peroxisome proliferator. Since there were no observed changes in lipid metabolism, this evidence suggests that enoyl-CoAs were diverted to the D-hydroxy-specific beta-oxidation system for metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3,2-trans-enoyl-CoA isomerase; enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase; enoyl-Coenzyme A, hydratase/3-hydroxyacyl Coenzyme A dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-bifunctional protein, peroxisomal; multifunctional enzyme 1; PBE; peroxisomal enoyl-CoA hydratase; unnamed protein product
Gene Aliases: ECHD; FRTS3; L-PBE; LBFP; MFE1; PBFE
UniProt ID: (Human) Q08426
Entrez Gene ID: (Human) 1962
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support