pUC57 DNA
Thermo Scientific™

pUC57 DNA

Thermo Scientific Plasmid pUC57, 2710 bp in length, is a derivative of pUC19. pUC57 MCS contains 6 restriction sites withRead more
Have Questions?
Catalog NumberQuantity
SD017150 μg
Catalog number SD0171
Price (USD)
111.00
50 µg
Quantity:
50 μg
Request bulk or custom format
Price (USD)
111.00
50 µg
Thermo Scientific Plasmid pUC57, 2710 bp in length, is a derivative of pUC19. pUC57 MCS contains 6 restriction sites with protruding 3’-ends that are resistant to E. coli exonuclease III. This vector is designed for cloning and generation of ExoIII deletions. The exact positions of the genetic elements are shown on the map (termination codons included). DNA replication initiates at position 890 (±1) and proceeds in the direction indicated. The bla gene nucleotides 2510-2442 (compl. strand) code for a signal peptide.

Highlights

• More than 90% in the supercoiled form
• Isolated from E. coli (dam+, dcm+)
• Purified by chromatography using proprietary patented technology

Applications

• Cloning
• Sequencing of insert DNA

Related Products
pUC18 DNA
pUC19 DNA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cloning MethodRestriction Enzyme⁄ MCS
Concentration0.5 μg/μL
Final Product TypeDNA
For Use With (Application)Cloning
Product TypeDNA
Quantity50 μg
Sample TypeDNA
VectorpUC57
Unit Size50 µg