SureLock™ Tandem Midi Gel Tank - FAQs
SureLock™ Tandem Midi Gel Tank - FAQs
View additional product information for SureLock™ Tandem Midi Gel Tank - FAQs (STM1001)
10 product FAQs found
Can I use a Bio-Rad™ power supply with the SureLock Tandem Midi Gel Tank (Cat. No. STM1001)?
If using a Bio-Rad™ power supply, Novex Power Supply Adapters (Cat. No. ZA10001) are NOT needed for the SureLock Tandem Midi Gel Tank and should be removed prior to use.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I use the PowerEase Touch 120W Power Supply to transfer two midi gels with the SureLock Tandem Midi Gel Tank and SureLock Tandem Midi Blot Module?
Unfortunately, the PowerEase Touch 120W power supply has a maximum output of 500 mA and is not suitable for transferring two midi gels. For wet transfer, we recommend using the PowerEase Touch Power Supply 350W or 600W unit, which can deliver up to 3 A. More information about these power supply units can be found on our website.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Can I use the PowerEase Touch 120W Power Supply with the SureLock Tandem Midi Gel Tank?
Yes, up to two midi gels can be run in the SureLock Tandem Midi Gel Tank using the PowerEase Touch 120W Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Which blot module is compatible with the SureLock Tandem Midi Gel Tank?
The SureLock Tandem Midi Blot Module (Cat. No. STM2001) is compatible with the SureLock Tandem Midi Gel Tank.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How many gels can I run using the SureLock Tandem Midi Gel Tank?
Up to two midi gels can be run at a time using the SureLock Tandem Midi Gel Tank, each in separate chambers.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
What are the power supply requirements for the SureLock Tandem Midi Gel Tank? Do you offer a power supply that I can use with it?
The SureLock Tandem Midi Gel Tank should be used with an external DC power supply designed for electrophoresis. This power supply must be isolated from the ground so that the DC output is floating (safety requirement) and must be equipped with No Load or Open Circuit protection. It must be able to operate at 300 V and have an output current minimum of at least 1 A (1000 mA). We recommend using the PowerEase Touch 350W Power Supply (Cat. No. PS0350) with the SureLock Tandem Midi Gel Tank. Note: To allow connection of the covered power leads of the SureLock Tandem Midi Gel Tank with certain power supplies that are not designed for use with covered or non-retractable power leads, we recommend using Novex Power Supply Adapters available separately (Cat. No. ZA10001).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
How do you recommend cleaning the SureLock Tandem Midi Gel Tank?
We recommend washing the SureLock Tandem Midi Gel Tank with a mild detergent and rinsing with deionized water after each use.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Is the SureLock Tandem Midi Gel Tank compatible with organic solvents?
The SureLock Tandem Midi Gel Tank is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene, etc.), acetone, ethyl alcohol, or isopropyl alcohol.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I run one gel using the SureLock Tandem Midi Gel Tank?
Yes, you can run 1 or 2 gels using the SureLock Tandem Midi Gel Tank. See the User Guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0018507_SureLockMidiGelTank_UG.pdf) for detailed instructions.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I purchase replacement parts for the SureLock Tandem Midi Gel Tank (Cat. No. STM1001)?
Yes, the following replacement parts are available for the SureLock Tandem Midi Gel Tank:
- SureLock Tandem Midi Gel Tank Replacement Lid, Cat. No. STM1002
- SureLock Tandem Midi Gel Tank Replacement Tank, Cat. No. STM1003
- SureLock Tandem Midi Gel Tank Replacement Cassette Clamp, Cat. No. STM1004
- Gel Knife, Cat. No. EI9010
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.