Search
View additional product information for SureLock™ Tandem Midi Gel Tank Replacement Lid - FAQs (STM1002)
6 product FAQs found
Unfortunately, the PowerEase Touch 120W power supply has a maximum output of 500 mA and is not suitable for transferring two midi gels. For wet transfer, we recommend using the PowerEase Touch Power Supply 350W or 600W unit, which can deliver up to 3 A. More information about these power supply units can be found on our website.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
No, most transfer applications require high current and therefore, we recommend using a high current power supply like PowerEase Touch 350W Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
We offer the following pre-cut membranes and filters for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, nitrocellulose, Cat. No. STM2008
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 µm, nitrocellulose, Cat. No. STM2007
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, PVDF, Cat. No. STM2006
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
The SureLock Tandem Midi Blot Module (Cat. No. STM2001) is compatible with the SureLock Tandem Midi Gel Tank.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Yes, the following replacement parts are available for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Gel Tank Replacement Tank, Cat. No. STM1003
- SureLock Tandem Midi Gel Tank Replacement Lid, Cat. No. STM1002
- SureLock Tandem Midi Gel Tank Replacement Cassette Clamp, Cat. No. STM1004
- Gel Knife, Cat. No. EI9010
- SureLock Tandem Midi Sponge Kit, Cat. No. STM2002
- Blotting Roller, Cat. No. LC2100
- SureLock Tandem Midi Blot Module Replacement Gasket Kit , Cat. No. STM2003
- SureLock Tandem Transfer Tray, Cat. No. STM3001
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Only one midi gel can be transferred at a time using the SureLock Tandem Midi Blot Module. The SureLock Tandem Mini Gel Tank accommodates one SureLock Tandem Midi Blot Module per chamber, or two blot modules total. Hence, upto two midi gels can be transferred at a time using one SureLock Tandem Mini Gel Tank fitted with two SureLock Tandem Midi Blot Modules.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.