SureLock™ Tandem Midi Gel Tank Replacement Lid
Invitrogen™
SureLock™ Tandem Midi Gel Tank Replacement Lid
This lid is a replacement part for the SureLock Tandem Midi Gel Tank.About the SureLock Tandem Midi Gel TankThe SureLockRead more
Have Questions?
Catalog NumberQuantity
STM10021 lid
Catalog number STM1002
Price (USD)
123.50
Special offer
Online exclusive
Ends: 27-Jun-2026
130.00
Save 6.50 (5%)
1 lid
Quantity:
1 lid
Price (USD)
123.50
Special offer
Online exclusive
Ends: 27-Jun-2026
130.00
Save 6.50 (5%)
1 lid
This lid is a replacement part for the SureLock Tandem Midi Gel Tank.
About the SureLock Tandem Midi Gel Tank
The SureLock Tandem Midi Gel Tank provides for rapid electrophoresis of midi gels using minimal buffer (∼520 mL/gel) in a leak-free system. With a set-up time of ∼30 seconds, the tank efficiently runs midi gels while also providing consistent performance. The tank features two independent chambers, allowing electrophoresis of one or two gels at a time, which saves on buffer and limits waste. The tank is compatible with all Invitrogen pre-cast midi gels and midi gel cassettes.
Specifications
Quantity1 lid
Shipping ConditionRoom Temperature
Warranty1 year
For Use With (Equipment)SureLock Tandem Midi Gel Tank
Gel SizeMidi
Product LineSureLock
TypeMidi Gel Tank Replacement Lid
Unit Size1 lid
Frequently asked questions (FAQs)
Can I use the PowerEase Touch 120W Power Supply to transfer two midi gels with the SureLock Tandem Midi Gel Tank and SureLock Tandem Midi Blot Module?
Unfortunately, the PowerEase Touch 120W power supply has a maximum output of 500 mA and is not suitable for transferring two midi gels. For wet transfer, we recommend using the PowerEase Touch Power Supply 350W or 600W unit, which can deliver up to 3 A. More information about these power supply units can be found on our website.
Find additional tips, troubleshooting help, and resources within our Protein Gel Electrophoresis Chambers, Power Supplies, and Accessories Support Center.
Can I use the PowerEase Touch 120W Power Supply for transferring gels with the SureLock Tandem Midi Blot Module, Mini Gel Blot Module, or Novex Semi-Dry Blotter?
No, most transfer applications require high current and therefore, we recommend using a high current power supply like PowerEase Touch 350W Power Supply.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Do you offer pre-cut membranes and filters for the SureLock Tandem Midi Blot Module?
We offer the following pre-cut membranes and filters for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, nitrocellulose, Cat. No. STM2008
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.2 µm, nitrocellulose, Cat. No. STM2007
- SureLock Tandem Midi Pre-cut Membranes and Filters, 0.45 µm, PVDF, Cat. No. STM2006
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Which blot module is compatible with the SureLock Tandem Midi Gel Tank?
The SureLock Tandem Midi Blot Module (Cat. No. STM2001) is compatible with the SureLock Tandem Midi Gel Tank.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Can I purchase replacement parts for the SureLock Tandem Midi Blot Module (Cat. No. STM2001)?
Yes, the following replacement parts are available for the SureLock Tandem Midi Blot Module:
- SureLock Tandem Midi Gel Tank Replacement Tank, Cat. No. STM1003
- SureLock Tandem Midi Gel Tank Replacement Lid, Cat. No. STM1002
- SureLock Tandem Midi Gel Tank Replacement Cassette Clamp, Cat. No. STM1004
- Gel Knife, Cat. No. EI9010
- SureLock Tandem Midi Sponge Kit, Cat. No. STM2002
- Blotting Roller, Cat. No. LC2100
- SureLock Tandem Midi Blot Module Replacement Gasket Kit , Cat. No. STM2003
- SureLock Tandem Transfer Tray, Cat. No. STM3001
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.