Product Image
Invitrogen™

pCEP4 Mammalian Expression Vector

These vectors are designed for high-level, constitutive expression from either the CMV or RSV promoters. Both vectors contain the EBNA-1Read more
Have Questions?
Catalog NumberQuantity
V0445020 μg
Catalog number V04450
Price (USD)
1,050.00
20 µg
Quantity:
20 μg
Price (USD)
1,050.00
20 µg
These vectors are designed for high-level, constitutive expression from either the CMV or RSV promoters. Both vectors contain the EBNA-1 gene for episomal expression in primate and canine cell lines.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Constitutive or Inducible SystemConstitutive
Delivery TypeTransfection
For Use With (Application)Stable Cell Line Development, Episomal Expression
Product TypeMammalian Expression Vector
Quantity20 μg
Selection Agent (Eukaryotic)Hygromycin
VectorpCEP4
Cloning MethodRestriction Enzyme/MCS
PromoterCMV
Protein TagUntagged
Unit Size20 µg
Contents & Storage
20 μg of pCEP4 vector in Tris-EDTA buffer

Store at -20°C. Vectors are guaranteed stable for 6 months when properly stored.

Frequently asked questions (FAQs)

How do your episomal mammalian expression vectors bring about episomal replication? Can I use these vectors in any cell line?

Our episomal mammalian expression vectors (pCEP4 and pREP4) contain the Epstein Barr Virus (EBV) origin of replication (oriP) and the Epstein-Barr nuclear antigen (EBNA-1) for high-copy, transient, or stable episomal replication in human, primate, canine, and porcine cell lines. They do not bring about episomal expression in murine or rodent cell lines.

I performed stable selection but my antibiotic-resistant clones do not express my gene of interest. What could have gone wrong?

Here are possible causes and solutions:

Detection method may not be appropriate or sensitive enough:
- We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot.
- Insufficient number of clones screened: Screen at least 20 clones.
- Inappropriate antibiotic concentration used for stable selection: Make sure the antibiotic kill curve was performed correctly. Since the potency of a given antibiotic depends upon cell type, serum, medium, and culture technique, the dose must be determined each time a stable selection is performed. Even the stable cell lines we offer may be more or less sensitive to the dose we recommend if the medium or serum is significantly different.
- Expression of gene product (even low level) may not be compatible with growth of the cell line: Use an inducible expression system.
- Negative clones may result from preferential linearization at a vector site critical for expression of the gene of interest: Linearize the vector at a site that is not critical for expression, such as within the bacterial resistance marker.

I used a mammalian expression vector but do not get any expression of my protein. Can you help me troubleshoot?

Here are possible causes and solutions:

- Try the control expression that is included in the kit
Possible detection problem:

- Detection of expressed protein may not be possible in a transient transfection, since the transfection efficiency may be too low for detection by methods that assess the entire transfected population. We recommend optimizing the transfection efficiency, doing stable selection, or using methods that permit examination of individual cells. You can also increase the level of expression by changing the promoter or cell type.
- Expression within the cell may be too low for the chosen detection method. We recommend optimizing the detection protocol or finding more sensitive methods. If the protein is being detected by Coomassie/silver staining, we recommend doing a western blot for increased sensitivity. The presence of endogenous proteins in the lysate may obscure the protein of interest in a Coomassie/silver stain. If available, we recommend using a positive control for the western blot. Protein might be degraded or truncated: Check on a Northern. Possible time-course issue: Since the expression of a protein over time will depend upon the nature of the protein, we always recommend doing a time course for expression. A pilot time-course assay will help to determine the optimal window for expression. Possible cloning issues: Verify clones by restriction digestion and/or sequencing.

Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.

I am using a mammalian expression vector that has the neomycin resistance gene. Can I use neomycin for stable selection in mammalian cells?

No; neomycin is toxic to mammalian cells. We recommend using Geneticin (a.k.a. G418 Sulfate), as it is a less toxic and very effective alternative for selection in mammalian cells.

Is it okay if my construct has an ATG that is upstream of the ATG in my gene of interest? Will it interfere with translation of my gene?

Translation initiation will occur at the first ATG encountered by the ribosome, although in the absence of a Kozak sequence, initiation will be relatively weak. Any insert downstream would express a fusion protein if it is in frame with this initial ATG, but levels of expressed protein are predicted to be low if there is a non-Kozak consensus sequence. If the vector contains a non-Kozak consensus ATG, we recommend that you clone your gene upstream of that ATG and include a Kozak sequence for optimal expression.

Citations & References (68)

Citations & References
Abstract
The gene for the embryonic stem cell coactivator UTF1 carries a regulatory element which selectively interacts with a complex composed of Oct-3/4 and Sox-2.
Authors:Nishimoto M,Fukushima A,Okuda A,Muramatsu M
Journal:Molecular and cellular biology
PubMed ID:10409735
UTF1 is a transcriptional coactivator which has recently been isolated and found to be expressed mainly in pluripotent embryonic stem (ES) cells (A. Okuda, A. Fukushima, M. Nishimoto, et al., EMBO J. 17:2019–2032, 1998). To gain insight into the regulatory network of gene expression in ES cells, we have characterized ... More
WAF1, a potential mediator of p53 tumor suppression.
Authors:el-Deiry WS, Tokino T, Velculescu VE, Levy DB, Parsons R, Trent JM, Lin D, Mercer WE, Kinzler KW, Vogelstein B
Journal:Cell
PubMed ID:8242752
The ability of p53 to activate transcription from specific sequences suggests that genes induced by p53 may mediate its biological role as a tumor suppressor. Using a subtractive hybridization approach, we identified a gene, named WAF1, whose induction was associated with wild- type but not mutant p53 gene expression in ... More
DNA methylation and genetic instability in colorectal cancer cells [see comments]
Authors:Lengauer C, Kinzler KW, Vogelstein B
Journal:Proc Natl Acad Sci U S A
PubMed ID:9122232
'Apparent alterations in DNA methylation have been observed in many cancers, but whether such alterations represent a persistent alteration in the normal methylation process is not known. In this study, we report a striking difference in the expression of exogenously introduced retroviral genes in various colorectal cancer cell lines. Extinguished ... More
Phosphorylation of glycogen synthase kinase-3 and stimulation of T-cell factor signaling following activation of EP2 and EP4 prostanoid receptors by prostaglandin E2.
Authors: Fujino Hiromichi; West Kimberly A; Regan John W;
Journal:J Biol Chem
PubMed ID:11706038
'Recently we have shown that the FP(B) prostanoid receptor, a G-protein-coupled receptor that couples to Galpha(q), activates T-cell factor (Tcf)/lymphoid enhancer factor (Lef)-mediated transcriptional activation (Fujino, H., and Regan, J. W. (2001) J. Biol. Chem. 276, 12489-12492). We now report that the EP(2) and EP(4) prostanoid receptors, which couple to ... More
The intramembrane cleavage site of the amyloid precursor protein depends on the length of its transmembrane domain.
Authors: Lichtenthaler Stefan F; Beher Dirk; Grimm Heike S; Wang Rong; Shearman Mark S; Masters Colin L; Beyreuther Konrad;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11805291
'Proteolytic processing of the amyloid precursor protein by beta-secretase generates C99, which subsequently is cleaved by gamma-secretase, yielding the amyloid beta peptide (A beta). This gamma-cleavage occurs within the transmembrane domain (TMD) of C99 and is similar to the intramembrane cleavage of Notch. However, Notch and C99 differ in their ... More