Orion™ Versa Star Pro™ pH/LogR™ Benchtop Meter
Thermo Scientific™

Orion™ Versa Star Pro™ pH/LogR™ Benchtop Meter

Get premium performance and ultimate flexibility with our benchtop meter with pH/LogR module for pH, mV, ORP and LogR temperature technology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
VSTAR80Meter, pH/LogR (Sensor Not Included)
VSTAR82Meter, pH/LogR, Difficult Sample Kit
VSTAR83Meter, pH/LogR, Micro Sample Kit
Catalog number VSTAR80
Price (USD)
2,421.00
Online exclusive
2,690.00
Save 269.00
Each
Type:
Meter, pH/LogR (Sensor Not Included)
Price (USD)
2,421.00
Online exclusive
2,690.00
Save 269.00
Each

Get high accuracy, premium performance and ultimate flexibility with the Thermo Scientific™ Orion™ Versa Star Pro™ pH/LogR™ Benchtop Meter. Meet your most challenging applications for pH, mV, ORP and LogR temperature. Easily view measurements on the stunning color display and customize the four meter channels with interchangeable modules. Use LogR technology to measure both pH and temperature using the glass-bulb of most pH electrodes. Log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer.

Thermo Scientific Orion Versa Star Pro Benchtop Meters are designed to meet your most challenging application with ease. Delivering rich functionality and features, you can optimally configure your meter system to meet changing requirements. The Orion Versa Star Pro Benchtop Meters offer interchangeable measurement modules that allow multiple users to customize four separate channels to meet their specific requirements. Easily view your measurements on a stunning color display with adjustable text sizes. Take the guesswork out of meter calibration and setup with onscreen instructions available in a variety of local languages. Enjoy quick and simple data entry using the numeric keypad.

  • Onscreen measurement stability indicator lets you know exactly when your measurements are ready
  • Selectable read modes include Auto-Read to hold a stable reading, Continuous to show changing readings, Timed to log data at specific intervals and Single-Shot to hold a reading after a specific wait time is reached
  • Audible alarms automatically notify you when a set offset value, high/low limit value and calibration time interval are reached
  • Easily access, update and switch your preferred measurement settings, using 10 password-protected methods per channel
  • Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position and can be quickly rinsed between samples
  • Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID and account ID
  • Calibration log stores 10 most recent calibrations per parameter with viewable graph for multipoint pH and ISE calibrations
  • Ensure measurement integrity by viewing the active calibration log while taking readings
  • Non-volatile meter memory preserves data and settings, even in the event of power loss
  • Meter facilitates data transfers to a computer via USB or RS232 connections
  • IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
  • Meters include CE, TUV 3-in-1 and FCC Class A certifications
  • 3-year replacement warranty

Includes Orion Versa Star pH/LogR Measurement Module:

  • pH, mV, relative mV or ORP with temperature using LogR technology, ATC probe or manual input
  • LogR technology uses the glass-bulb of most pH electrodes to measure both pH and temperature at the same location — ideal for small sample volumes and samples that are difficult to clean
  • Perform up to a 6 point pH calibration using automatic buffer recognition of NIST, DIN or user-defined buffer groups with choice to manually enter custom buffer values
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading

System Access Feature
Orion Versa Star Pro Meters offer a system access feature that is designed to help ensure data integrity by preventing undesired access to various meter functions including date/time setting, data log and calibration log. The System Access feature can be easily turned on or off as the needs of your lab change for ultimate flexibility in operation and ease-of-use. When enabled, the System Access feature allows assignment of a system administrator and up to ten user accounts with customizable access to instrument settings, measurement settings, calibrations and logs. Use the password protection to secure selected meter functions, while leaving other functions accessible for all users. The System Access feature allows you to customize meter access to meet your specific security and data reporting needs.

Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Specifications
AC AdapterUniversal 50-60 Hz, 100-240 VAC power adapter (Cat. No. STARA-PWR)
Accuracy (Temperature)±0.1°C
Accuracy (mV)±0.2 or ±0.05%, whichever is greater
Accuracy (pH)±0.002
Alarm OutputSet point alarm, high/low limit alarm, calibration due alarm
Backlight OptionYes
Certifications/ComplianceCE, TUV 3-1, FCC Class A
Channels4 (Customizable with modules)
Data Management DetailsTransfer single, range or all data points to printer or computer
DescriptionOrion Versa Star Pro pH/LogR benchtop meter with stand
Dimensions (L x W x H)28 cm x 24 cm x 10 cm (11 in. x 9.5 in. x 3.9 in.), modules add ≤ 2 cm (0.8 in.) L & H
IP RatingIP54
IncludesVersa Star Pro meter with pH/LogR module, electrode stand, 100-240V universal power adapter, computer cable
InputsBNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (ATC temperature probe), two stirrer inputs (stirrer probe)
Isopotential Point7.000pH (adjustable)
KeypadNumeric with menu-specific function keys and dual purpose scroll/shortcut keys
LanguagesEnglish, Spanish, French, Italian, German, Chinese, Portuguese, Korean
MemoryNon-volatile memory preserves data log, calibration log and meter settings
Methods10 per channel
OutputsUSB, RS-232
Power RatingDC input: 9VDC, 1A
Power SupplyUniversal AC adapter (included)
Probe TypepH electrode, ORP electrode, reference half-cell electrode, ATC temperature probe, stirrer probe (purchase separately)
Range (ORP)±2000.0mV
Range (Relative mV)±2000.0mV
Relative Humidity Range5 to 85%, non-condensing
Resolution (Temperature)0.1°C, 0.1°F
Resolution (mV)0.1mV
StabilitySelectable fast, medium, slow or Smart Stability setting; Selectable off or Automatic Smart averaging setting
Temperature (English) Operating41°F to 113°F (ambient)
Temperature (Metric) Operating5°C to 45°C (ambient)
Temperature Probe Calibration1 point temperature offset calibration, 3 point LogR temperature calibration
Temperature Range (English)22.0°F to 221.0°F
Temperature SelectionManual or automatic with ATC temperature probe, automatic with pH electrode using LogR mode
TypeMeter, pH/LogR (Sensor Not Included)
Warranty3 Year Meter and Module Warranty
Weight (English)2.5 lb., modules add 0.5 lb. each
Weight (Metric)1.2 kg, modules add 0.2 kg each
CalibrationpH with calibration editing option, relative mV, ORP, LogR temperature and temperature
Calibration Points1 to 6
Data Points2000 with Date and Time Stamp
Display TypeLCD, Color Graphic with User-Customizable Options
Log Function TypeAutomatic with Auto-Read, Timed, and Single-shot Measure Modes, Manual
Measurement ModespH with LogR Temperature, mV, Relative mV, ORP with Temperature
Range (mV)±2000.0
Range (pH)-2 to 20
Resolution (pH)0.001, 0.01, 0.1
Temperature Range (Metric)-5°C to +105°C
Unit SizeEach

Frequently asked questions (FAQs)

How long is the warranty period for Orion pH meters?

At Thermo Fisher Scientific, we stand behind the quality and reliability of our instruments. That's why we offer a minimum two-year (2) warranty on all Orion benchtop and portable pH meters. Obtain peace of mind knowing that your investment is protected. Should any issues arise within the warranty period, our team of experts is committed to providing you with prompt and efficient service. Be sure to check instrument specifications to confirm the warranty coverage period.

How do I perform a calibration using Orion pH meters?

Calibration is an important step to obtaining accurate results on your Orion benchtop or portable pH meter. Before beginning the calibration process, ensure that you are utilizing freshly poured, non-expired calibration buffers to help ensure accurate results. Follow the steps below to calibrate your pH meter.

• Prepare the calibration solutions: Obtain at least two pH reference solutions with known pH values. Ideally, use one solution close to the expected pH of your sample, and another solution at a different pH value. Ensure that the solutions are fresh and have not expired.
• Clean the electrode: Rinse the electrode with distilled or deionized water to remove any debris or contaminants. If necessary, gently wipe the electrode with a soft cloth or tissue.
• Turn on the pH meter: Power on the pH meter and allow it to warm up, if needed.
• Immerse the electrode: Immerse the electrode into a beaker containing the first calibration solution. Ensure that the electrode is fully submerged and that it is not touching the sides or bottom of the beaker.
• Allow stabilization: Allow the pH reading to stabilize. This may take a few moments, depending on the specific meter and solution being used. The reading should become steady and remain constant.
• Adjust the pH value: Using the calibration controls on the pH meter, adjust the reading to match the known pH value of the calibration solution. Reference the user manual for specific guidance on adjusting the pH.
• Rinse the electrode: After calibration, rinse the electrode with distilled or deionized water to remove any residual calibration solution.
• Repeat calibration: Repeat the calibration steps with the second calibration solution, following the same process as before.
• Verify accuracy: After calibration, it is recommended to check the pH meter's accuracy by measuring a pH solution of known value. This can help ensure the reliability of your measurements.

To learn more about pH meter calibration, download our technical note: Understanding Calibration of Laboratory pH Systems. See a live calibration by watching our video: Understanding Calibration of Laboratory pH Systems.

How do I perform an analysis using Orion pH meters?

Before beginning an analysis on your Orion benchtop or portable pH meter, assemble your materials. You will need a pH electrode suitable for your application, calibration buffer solutions, and a test sample. Follow the steps below to obtain your pH analysis.

• Calibrate the pH meter: Start by calibrating your pH meter using the calibration buffer solutions. Typically, you will need pH 4, pH 7, and pH 10. Be sure to reference the user manual for instructions on how to properly calibrate your pH meter.
• Prepare your sample: Ensure that your sample is at the desired temperature (most frequently room temperature). If necessary, filter the sample to remove any particles or impurities that could impact the pH reading.
• Immerse the electrode: Dip the pH electrode into the sample. Avoid touching the sides or bottom of the container to prevent inaccurate readings.
• Allow for stabilization: Give the pH meter some time to stabilize and reach a steady reading. This may take a few seconds to a minute depending on your meter and the characteristics of the sample.
• Record the pH value: Once the pH meter stabilizes, record the pH value displayed on the meter. If applicable, also record the temperature value.
• Rinse the electrode: After each measurement, rinse the electrode with distilled or deionized water to remove any residue or contaminants. This helps ensure accurate subsequent readings.
• Repeat (if necessary): If you need to analyze multiple samples, repeat the process with each sample, recalibrating the pH meter if required.

To learn more about how to perform a pH analysis, watch our video:Measuring Drinking Water Using the Orion Versa Star Pro Bench Meter

How do I choose an Orion pH electrode for my application?

When choosing an Orion pH electrode for your application there are many factors to consider. Sample type, container dimensions, accuracy/ precision requirements, junction design, durability, maintenance, and meter compatibility are all key features to evaluate.
To learn more about how to choose the right electrode for your application, watch our videos: How to Choose the Right pH Electrode and Understanding Your Sample to Select the Right pH Electrode

What supplies do I need to perform an analysis with Orion pH meters?

To perform an analysis using Orion benchtop and portable pH meters, you will need the following supplies: pH electrode (or micro electrode for small sample volumes/ containers), calibration buffer solutions, distilled or deionized water, sample containers, stirrer probe (optional), and electrode cleaning solution(s) (s). For your convenience, our all-in-one pH buffer kits can be purchased, which include pH buffers (4.01 pH, 7.00 pH, and 10.01 pH), electrode cleaning solution, and electrode and storage solution.