Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
A more sustainable equivalent is available.
Tubes and Domed Caps, strips of 8
Thermo Scientific™

Tubes and Domed Caps, strips of 8

Thermo Scientific Tubes and Domed Caps, Strips of 8 are designed for PCR applications, and are suitable for 0.2 mLRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct TypeQuantity
AB0451Natural8-Strip Tube10 Packs of 12 Strips
AB0266Natural8-Strip Tube250 Strips
Catalog number AB0451
Price (USD)
208.00
Each
Color:
Natural
Product Type:
8-Strip Tube
Quantity:
10 Packs of 12 Strips
Request bulk or custom format
Price (USD)
208.00
Each
Thermo Scientific Tubes and Domed Caps, Strips of 8 are designed for PCR applications, and are suitable for 0.2 mL thermal cycler blocks and 96-well V-bottom thermal cyclers. The 8-strip tubes can be divided into individual tubes.

Features of Thermo Scientific Tubes and Domed Caps strips:

• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when capped
• 8 tube strips can be cut into individual tubes
• Ideal for use in 96-well V-bottom thermal cyclers

Applications

• PCR

Related Products
Tubes and Domed Caps, strips of 8, blue
Tubes and Domed Caps, strips of 8, green
Tubes and Domed Caps, strips of 8, red
Tubes and Domed Caps, strips of 8, yellow
Tubes and Domed Caps, strips of 8
Tubes and Flat Caps, strips of 8
Tubes and Flat Caps, strips of 8, assorted colors
Tubes and Ultra Clear Caps, strips of 8
Tubes (white) and Ultra Clear Caps, strips of 8
Tubes and Domed Caps, strips of 12
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers
Format120 Strips Tubes and Caps
No. of Wells96
Quantity10 Packs of 12 Strips
Reaction SpeedStandard
Volume (Metric) Working0.2 mL
Closure TypeDomed Cap
ColorNatural
MaterialPolypropylene
Product Type8-Strip Tube
Volume (Metric)0.25 mL
Unit SizeEach