Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Description: The antibody reacts with mouse lymphotoxin-beta receptor (LTBR). Northern blot analysis of tissues from adult mice showed that expression levels of LTBR mRNA were strong in lung, liver, and kidney, moderate in heart and testes, but weak in brain, thymus, spleen, and lymph nodes. The tumor necrosis factor receptor-related protein is the human receptor for the heterotrimer of lymphotoxin-alpha and lymphotoxin-beta. This LT-alpha/LT-beta heterotrimer (LT-alpha1beta2) is assumed to take part in immunologic reactions by cell-cell contact, but does not bind to either TNFR1 (CD120alpha) or TNFR2 (CD120beta). LTBR is expressed by day 7 embryos, so it has been speculated that the LT-alpha/LT-beta receptor system may also have some function in early embryogenesis.
Applications Reported: This eBio3C8 (3C8) antibody has been reported for use in flow cytometric analysis.
Applications Tested: This eBio3C8 (3C8) antibody has been tested by flow cytometric analysis of the NIH/3T3 cell line. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10e5 to 10e8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
Filtration: 0.2 µm post-manufacturing filtered.
The protein encoded by this gene is a member of the tumor necrosis factor (TNF) family of receptors. It is expressed on the surface of most cell types, including cells of epithelial and myeloid lineages, but not on T and B lymphocytes. The protein specifically binds the lymphotoxin membrane form (a complex of lymphotoxin-alpha and lymphtoxin-beta). The encoded protein and its ligand play a role in the development and organization of lymphoid tissue and tranformed cells. Activation of the encoded protein can trigger apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LT beta-R; LT-beta receptor; Lymphotoxin-beta receptor; TNF receptor-related protein; TNFR-III; Tumor necrosis factor receptor superfamily member 3; tumor necrosis factor receptor superfamily, member 3
Gene Aliases: AI256028; CD18; Ltar; LTbetaR; Ltbr; TNF-R-III; Tnfbr; TNFCR; TNFR-RP; TNFR2-RP; TNFRrp; Tnfrsf3
UniProt ID: (Mouse) P50284
Entrez Gene ID: (Mouse) 17000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support