Product References
Single-cell RNA Sequencing Identified Novel Nr4a1+ Ear2+ Anti-Inflammatory Macrophage Phenotype under Myeloid-TLR4 Dependent Regulation in Anti-Glomerular Basement Membrane (GBM) Crescentic Glomerulonephritis (cGN).
Advanced science (Weinheim, Baden-Wurttemberg, Germany)
Chen J,Huang XR,Yang F,Yiu WH,Yu X,Tang SCW,Lan HY
12-5965-82 was used in flow cytometry to study single-cell RNA sequencing (scRNA-seq), pseudotime trajectories reconstruction, and motif enrichment analysis, and macrophage diversity in anti-GBM cGN under tight regulation of myeloid-TLR4 to decode a previously unidentified role for a myeloid-TLR4 dependent Nr4a1/Ear2 negative feedback mechanism in macrophage-mediated progressive renal injury, implying that activation of Nr4a1-Ear2 axis can be a novel and effective immunotherapy for anti-GBM cGN.
Wed Jun 01 00:00:00 EDT 2022
MTHFD2 is a metabolic checkpoint controlling effector and regulatory T cell fate and function.
Immunity
Sugiura A,Andrejeva G,Voss K,Heintzman DR,Xu X,Madden MZ,Ye X,Beier KL,Chowdhury NU,Wolf MM,Young AC,Greenwood DL,Sewell AE,Shahi SK,Freedman SN,Cameron AM,Foerch P,Bourne T,Garcia-Canaveras JC,Karijolich J,Newcomb DC,Mangalam AK,Rabinowitz JD,Rathmell JC
12-5965-82 was used in Flow cytometry/Cell sorting to show that MTHFD2 is a metabolic checkpoint to integrate purine metabolism with pathogenic effector cell signaling and is a potential therapeutic target within 1C metabolism pathways.
Tue Jan 11 00:00:00 EST 2022
Development of a B-cell maturation antigen-specific T-cell antigen coupler receptor for multiple myeloma.
Cytotherapy
Bezverbnaya K,Moogk D,Cummings D,Baker CL,Aarts C,Denisova G,Sun M,McNicol JD,Turner RC,Rullo AF,Foley SR,Bramson JL
12-5965-82 was used in Flow cytometry/Cell sorting to represent an important step in the evolution of this technology, which tolerates a range of expression levels without impacting therapeutic efficacy.
Wed Sep 01 00:00:00 EDT 2021
T cell self-reactivity during thymic development dictates the timing of positive selection.
eLife
Lutes LK,Steier Z,McIntyre LL,Pandey S,Kaminski J,Hoover AR,Ariotti S,Streets A,Yosef N,Robey EA
12-5965-82 was used in Flow Cytometry to imply that thymocytes with low self-reactivity downregulate TCR sensitivity more slowly during positive selection, and associate membrane ion channel expression with thymocyte self-reactivity and progress through positive selection.
Thu Apr 22 00:00:00 EDT 2021
T cell self-reactivity during thymic development dictates the timing of positive selection.
eLife
Lutes LK,Steier Z,McIntyre LL,Pandey S,Kaminski J,Hoover AR,Ariotti S,Streets A,Yosef N,Robey EA
12-5965-82 was used in Flow Cytometry to imply that thymocytes with low self-reactivity downregulate TCR sensitivity more slowly during positive selection, and associate membrane ion channel expression with thymocyte self-reactivity and progress through positive selection.
Thu Apr 22 00:00:00 EDT 2021
An Integrated Epigenomic and Transcriptomic Map of Mouse and Human αβ T Cell Development.
Immunity
Chopp LB,Gopalan V,Ciucci T,Ruchinskas A,Rae Z,Lagarde M,Gao Y,Li C,Bosticardo M,Pala F,Livak F,Kelly MC,Hannenhalli S,Bosselut R
12-5965-82 was used in Flow Cytometry to identify mouse and human αβ thymocyte developmental trajectories.
Tue Dec 15 00:00:00 EST 2020
An Integrated Epigenomic and Transcriptomic Map of Mouse and Human αβ T Cell Development.
Immunity
Chopp LB,Gopalan V,Ciucci T,Ruchinskas A,Rae Z,Lagarde M,Gao Y,Li C,Bosticardo M,Pala F,Livak F,Kelly MC,Hannenhalli S,Bosselut R
12-5965-82 was used in Flow Cytometry to identify mouse and human αβ thymocyte developmental trajectories.
Tue Dec 15 00:00:00 EST 2020
Dynamic regulation of chromatin organizer SATB1 via TCR-induced alternative promoter switch during T-cell development.
Nucleic acids research
Patta I,Madhok A,Khare S,Gottimukkala KP,Verma A,Giri S,Dandewad V,Seshadri V,Lal G,Misra-Sen J,Galande S
12-5965 was used in Flow cytometry/Cell sorting to study the mechanism regulating expression of SATB1 during T-cell development.
Fri Jun 19 00:00:00 EDT 2020
Dynamic regulation of chromatin organizer SATB1 via TCR-induced alternative promoter switch during T-cell development.
Nucleic acids research
Patta I,Madhok A,Khare S,Gottimukkala KP,Verma A,Giri S,Dandewad V,Seshadri V,Lal G,Misra-Sen J,Galande S
12-5965 was used in Flow cytometry/Cell sorting to study the mechanism regulating expression of SATB1 during T-cell development.
Fri Jun 19 00:00:00 EDT 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
12-5965 was used in Flow cytometry/Cell sorting to elucidate features of of human prenatal skin lymphocytes by using mass and flow cytometry.
Tue Nov 17 00:00:00 EST 2020
Developing Human Skin Contains Lymphocytes Demonstrating a Memory Signature.
Cell reports. Medicine
Dhariwala MO,Karthikeyan D,Vasquez KS,Farhat S,Weckel A,Taravati K,Leitner EG,Clancy S,Pauli M,Piper ML,Cohen JN,Ashouri JF,Lowe MM,Rosenblum MD,Scharschmidt TC
12-5965 was used in Flow cytometry/Cell sorting to elucidate features of of human prenatal skin lymphocytes by using mass and flow cytometry.
Tue Nov 17 00:00:00 EST 2020
Chronic Inflammation Permanently Reshapes Tissue-Resident Immunity in Celiac Disease.
Cell
Mayassi T,Ladell K,Gudjonson H,McLaren JE,Shaw DG,Tran MT,Rokicka JJ,Lawrence I,Grenier JC,van Unen V,Ciszewski C,Dimaano M,Sayegh HE,Kumar V,Wijmenga C,Green PHR,Gokhale R,Jericho H,Semrad CE,Guandalini S,Dinner AR,Kupfer SS,Reid HH,Barreiro LB,Rossjohn J,Price DA,Jabri B
12-5965-82 was used in Flow Cytometry to determine the role of tissue-resident lymphocytes in chronic inflammation.
Thu Feb 21 00:00:00 EST 2019
A Thpok-Directed Transcriptional Circuitry Promotes Bcl6 and Maf Expression to Orchestrate T Follicular Helper Differentiation.
Immunity
Vacchio MS,Ciucci T,Gao Y,Watanabe M,Balmaceno-Criss M,McGinty MT,Huang A,Xiao Q,McConkey C,Zhao Y,Shetty J,Tran B,Pepper M,Vahedi G,Jenkins MK,McGavern DB,Bosselut R
12-5965-82 was used in Flow Cytometry to identify a transcriptional program that links the CD4+ lineage with Tfh differentiation, a limiting factor for efficient B cell responses.
Tue Sep 17 00:00:00 EDT 2019
CD161 contributes to prenatal immune suppression of IFNγ-producing PLZF+ T cells.
The Journal of clinical investigation
Halkias J,Rackaityte E,Hillman SL,Aran D,Mendoza VF,Marshall LR,MacKenzie TC,Burt TD
Published figure using Nur77 monoclonal antibody (Product # 12-5965-82) in Flow Cytometry
Thu May 30 00:00:00 EDT 2019
CD161 contributes to prenatal immune suppression of IFNγ-producing PLZF+ T cells.
The Journal of clinical investigation
Halkias J,Rackaityte E,Hillman SL,Aran D,Mendoza VF,Marshall LR,MacKenzie TC,Burt TD
Published figure using Nur77 monoclonal antibody (Product # 12-5965-82) in Flow Cytometry
Thu May 30 00:00:00 EDT 2019
Restricted Expression of the Thymoproteasome Is Required for Thymic Selection and Peripheral Homeostasis of CD8+ T Cells.
Cell reports
Tomaru U,Konno S,Miyajima S,Kimoto R,Onodera M,Kiuchi S,Murata S,Ishizu A,Kasahara M
12-5965 was used in Flow cytometry/Cell sorting to find that thymoproteasomes are essential for positive selection but that the subsequent change in peptide repertoire in the medulla is also crucial for thymic selection and that β5t-derived peptide must be confined to the thymus to avoid autoimmunity in peripheral tissues.
Tue Jan 15 00:00:00 EST 2019
IgM and IgD B cell receptors differentially respond to endogenous antigens and control B cell fate.
eLife
Noviski M,Mueller JL,Satterthwaite A,Garrett-Sinha LA,Brombacher F,Zikherman J
12-5965 was used in Flow cytometry/Cell sorting to study how IgM and IgD B cell receptors differentially respond to endogenous antigens and control B cell fate.
Fri Mar 09 00:00:00 EST 2018
2D Kinetic Analysis of TCR and CD8 Coreceptor for LCMV GP33 Epitopes.
Frontiers in immunology
Kolawole EM,Andargachew R,Liu B,Jacobs JR,Evavold BD
12-5965 was used in Flow cytometry/Cell sorting to study TCR interaction in closely related GP33 variants.
Mon Sep 30 00:00:00 EDT 2019
Capacity of tTreg generation is not impaired in the atrophied thymus.
PLoS biology
Oh J,Wang W,Thomas R,Su DM
Wed Nov 01 00:00:00 EDT 2017
Calcium-mediated shaping of naive CD4 T-cell phenotype and function.
eLife
Guichard V,Bonilla N,Durand A,Audemard-Verger A,Guilbert T,Martin B,Lucas B,Auffray C
12-5965 was used in Flow cytometry/Cell sorting to demonstrate that calcium-mediated activation of the calcineurin pathway acts as a rheostat to shape the phenotype and effector potential of naive CD4 T cells in the steady-state.
Thu Dec 14 00:00:00 EST 2017
Thymic regulatory T cell niche size is dictated by limiting IL-2 from antigen-bearing dendritic cells and feedback competition.
Nature immunology
Weist BM,Kurd N,Boussier J,Chan SW,Robey EA
12-5965 was used in Flow cytometry/Cell sorting to investigate the sources of thymic IL-2 that limit access to the Treg developmental niche, showing that niche size is dictated by limiting IL-2 from antigen-bearing dendritic cells and feedback competition.
Mon Jun 01 00:00:00 EDT 2015
A sharp T-cell antigen receptor signaling threshold for T-cell proliferation.
Proceedings of the National Academy of Sciences of the United States of America
Au-Yeung BB,Zikherman J,Mueller JL,Ashouri JF,Matloubian M,Cheng DA,Chen Y,Shokat KM,Weiss A
12-5965 was used in Flow cytometry/Cell sorting to reveal a sharp threshold for the magnitude and duration of T-cell antigen receptor signalling required for commitment of T cells to proliferation.
Tue Sep 02 00:00:00 EDT 2014
Reduced TCR signaling potential impairs negative selection but does not result in autoimmune disease.
The Journal of experimental medicine
Hwang S,Song KD,Lesourne R,Lee J,Pinkhasov J,Li L,El-Khoury D,Love PE
Mon Sep 24 00:00:00 EDT 2012